Skip to Content
Merck
All Photos(1)

Key Documents

SAB4700374

Sigma-Aldrich

Monoclonal Anti-CD41a antibody produced in mouse

clone HIP8, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-ITGA2B

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$345.00

$345.00


Check Cart for Availability


Select a Size

Change View
100 μG
$345.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$345.00


Check Cart for Availability

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

HIP8, monoclonal

form

buffered aqueous solution

species reactivity

nonhuman primates, human

concentration

1 mg/mL

technique(s)

flow cytometry: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... ITGA2B(3674)

General description

Cluster of differentiation 41 (CD41), also known as integrin subunit α 2b (ITGA2B) or GPIIb/IIIa (glycoprotein), is encoded by the gene mapped to human chromosome 17q21.31. It is a platelet-specific, surface membrane receptor.
The antibody HIP8 reacts with alpha (α) subunit of CD41 (heavy chain; 120 kDa). CD41 is mainly expressed on platelets and megakaryocytes. The antibody HIP8 blocks platelet aggregation and completely inhibits ADP-, epinephrine-, and collagen-induced platelet activation, and partially inhibits ristocetin- and thrombin-induced platelet activation. HIP8 is useful in the morphological and physiological studies of platelets and megakaryocytes.

Application

The reagent is designed for Flow Cytometry analysis of human blood cells using 0.5 μg reagent / 1e6 cells in a suspension

Biochem/physiol Actions

Cluster of differentiation 41(CD41) is significantly involved in platelet adhesion and thrombus formation at the location of vascular injury. Fibrinogen and many extracellular matrix proteins bind in a groove between the αand β-subunits of the integrin receptor. Platelet aggregation, induced by the binding of fibrinogen to the ITGA2B receptor, is involved in the pathogenesis of acute thrombosis in coronary heart disease, stroke and peripheral arterial disease. CD41, expressed in haematopoietic progenitors, is associated with erythroid activity and endothelial marker expression. Single-nucleotide polymorphisms (SNPs) in CD41 might be associated with the risk of acute coronary syndrome and atherosclerosis.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline, pH 7.4, with 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Identification of ITGA2B and ITGB3 Single-Nucleotide Polymorphisms and Their Influences on the Platelet Function.
Xiang Q
BioMed Research International (2016)
CD41 and CD45 expression marks the angioformative initiation of neovascularisation in human haemangioblastoma.
Ma D
Tumour Biology : the Journal of the International Society For Oncodevelopmental Biology and Medicine, 37, 3765-3774 (2016)
β-Subunit Binding Is Sufficient for Ligands to Open the Integrin αIIbβ3 Headpiece.
Lin FY
The Journal of Biological Chemistry, 291, 4537-4546 (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service