Skip to Content
Merck
All Photos(1)

Key Documents

SAB4700278

Sigma-Aldrich

Monoclonal Anti-CD16-FITC antibody produced in mouse

clone MEM-154, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-FCGR3A, Anti-FcγRIII

Sign Into View Organizational & Contract Pricing

Select a Size

100 TEST
$377.00

$377.00


Estimated to ship on10 May 2025



Select a Size

Change View
100 TEST
$377.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.44

$377.00


Estimated to ship on10 May 2025


biological source

mouse

Quality Level

conjugate

FITC conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

MEM-154, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

flow cytometry: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... FCGR3A(2214)

Related Categories

General description

The antibody MEM-154 reacts with the epitope on CD16 antigen that residing in proximity to FG loop (probably BC or C′E loop). CD16 is a low affinity receptor for aggregated IgG (FcgammaRIII antigen). The antibody MEM-154 reacts with CD16+ granulocytes.

Immunogen

Human granulocytes

Application

The reagent is designed for Flow Cytometry analysis of human blood cells using 20 μL reagent / 100 μL of whole blood or 1e6 cells in a suspension. The content of a vial (2 mL) is sufficient for 100 tests.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline containing 15 mM sodium azide and 0.2% high-grade protease free BSA as a stabilizing agent.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service