Skip to Content
Merck
All Photos(1)

Key Documents

SAB4600461

Sigma-Aldrich

Anti-Rabbit IgG (H+L), highly cross-adsorbed, CF405M antibody produced in goat

~2 mg/mL, affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing

Select a Size

250 μL
$364.00

$364.00


Check Cart for Availability


Select a Size

Change View
250 μL
$364.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

$364.00


Check Cart for Availability

biological source

goat

Quality Level

conjugate

CF 405M conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

rabbit

concentration

~2 mg/mL

technique(s)

flow cytometry: 1-10 μg/mL
immunocytochemistry: suitable
immunohistochemistry: suitable
indirect ELISA: suitable
indirect immunofluorescence: 1-10 μg/mL
western blot: suitable

fluorescence

λex 408 nm; λem 452 nm

storage temp.

−20°C

target post-translational modification

unmodified

General description

Anti-Rabbit IgG (H+L) is a secondary antibody raised in goat against primary rabbit immunoglobulin G (IgG). The monomeric antibody is a ′Y′ shaped structure with two light chains and two heavy chains connected by disulfide bonds in the conserved cysteine residues. Both the light and heavy chains possess variable and constant domain. Light chain has one variable and one constant domain and heavy chain has one variable and three constant domains. IgG is about 150 kDa and is the predominant immunoglobulin in the body. The half-life of IgG is longest compared to other types of immunoglobulins. In bovidae, there are two major subtypes of IgG, namely IgG1 and IgG2. IgG1 is the predominant IgG in serum and colostrum.
This product is prepared by labeling highly cross-adsorbed goat anti-rabbit IgG (H+L) with CF405M. CF405M is spectrally similar to and as bright as Pacific Blue dye but is significantly more photostable than the latter. An additional advantage of CF405M is that it has less spillover in the 525/50 green channel than Pacific Blue dye due to narrower emission peak, making CF405M a superior choice in multi-color detection applications. To minimize cross-reactivity, the antibody has been adsorbed against Human, Mouse, and Rat serum. 

Specificity

Binds all rabbit IgGs (minimal cross-reaction with human, mouse, rat serum proteins)

Immunogen

rabbit IgG(H+L)

Biochem/physiol Actions

Immunoglobulins (Ig) has a wide range of antigenic determinant regions also known as paratope region that binds specific antigen and elicits immune response. The specificity is contributed by the amino acid sequences in the variable region of both light and heavy chains encoded by V genes. Other regions of the antibody has the constant regions encoded by C genes which accounts for its physiological functions.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied in phosphate buffered saline with 0.05% sodium azide, 50% glycerol and 0.2% BSA.

Preparation Note

Protect from light. The antibody solution should be gently mixed before use.

Legal Information

This product is distributed by Sigma-Aldrich Co. under the authorization of Biotium, Inc. This product is covered by one or more US patents and corresponding patent claims outside the US patents or pending applications owned or licensed by Biotium, Inc. including without limitation: 12/334,387; 12/607,915; 12/699,778; 12/850,578; 61/454,484. In consideration of the purchase price paid by the buyer, the buyer is hereby granted a limited, non-exclusive, non-transferable license to use only the purchased amount of the product solely for the buyer′s own internal research in a manner consistent with the accompanying product literature. Except as expressly granted herein, the sale of this product does not grant to or convey upon the buyer any license, expressly, by implication or estoppel, under any patent right or other intellectual property right of Biotium, Inc. Buyer shall not resell or transfer this product to any third party, or use the product for any commercial purposes, including without limitation, any diagnostic, therapeutic or prophylactic uses. This product is for research use only. Any other uses, including diagnostic uses, require a separate license from Biotium, Inc. For information on purchasing a license to use this product for purposes other than research, contact Biotium, Inc., 3159 Corporate Place, Hayward, CA 94545, Tel: (510) 265-1027. Fax: (510) 265-1352. Email: [email protected].
CF is a trademark of Biotium, Inc.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The covalent structure of an entire gamma G immunoglobulin molecule
Edelman GM, et al.
Proceedings of the National Academy of Sciences of the USA, 63(1), 78-85 (1969)
Biological properties of goat immunoglobulins G
Micusan VV and Borduas AG
Immunology, 32(4), 373-381 (1977)
Raul Lagos-Cabré et al.
Cell reports, 33(11), 108483-108483 (2020-12-17)
The mitotic spindle distributes chromosomes evenly to daughter cells during mitosis. The orientation of the spindle, guided by internal and external cues, determines the axis of cell division and thereby contributes to tissue morphogenesis. Progression through mitosis requires local Ca2+
Structure and function of immunoglobulins.
Schroeder Jr H W and Cavacini L
The Journal of Allergy and Clinical Immunology, 125(2), S41-S52 (2010)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service