Skip to Content
Merck
All Photos(8)

Key Documents

SAB4504450

Sigma-Aldrich

Anti-phospho-SAPK/JNK (pTyr185) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-JNK, Anti-JNK-46, Anti-JNK1, Anti-JNK1A2, Anti-JNK21B1/2, Anti-PRKM8, Anti-SAPK1, Anti-SAPK1c

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$529.00

$529.00


Check Cart for Availability


Select a Size

Change View
100 μG
$529.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$529.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 48 kDa

species reactivity

mouse, rat, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:20000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pTyr185)

Gene Information

General description

MAPK9 (mitogen-activated protein kinase 9)/JNK2 (C-Jun N-terminal kinase 2)/ stress-activated protein kinase 2 (SAPK2) spans around 58494bp. The JNK2 gene is located on human chromosome 5q35. It has 17 introns.

Immunogen

The antiserum was produced against synthesized peptide derived from human SAPK/JNK around the phosphorylation site of Tyr185.

Immunogen Range: 151-200

Biochem/physiol Actions

Stress-activated protein kinase 2 (SAPK2) participates in proliferation, differentiation, apoptosis, migration, transcriptional regulation and development. It controls the cellular acidic/lysosomal cellular compartment. JNK2 plays an important role in mediating stress-induced apoptosis and modulating gene expression.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

JNK2 is activated during ER stress and promotes cell survival
Raciti M, et al.
Cell Death & Disease (2012)
MAPK9 (mitogen-activated protein kinase 9)
Chen F
Atlas of Genetics and Cytogenetics in Oncology and Haematology (2003)
Junhui Liu et al.
Annals of translational medicine, 9(3), 260-260 (2021-03-13)
Kiaa0101, a regulator of cell proliferation, is overexpressed in many malignant tumors. However, its role in promoting invasion of glioma is poorly understood. Here, we investigated the effects of Kiaa0101 on glioma invasion and elucidated the underlying mechanisms of action.
Chin-Feng Hsuan et al.
Molecules (Basel, Switzerland), 25(24) (2020-12-17)
The proliferation and migration of vascular smooth muscle cells (VSMCs) are essential in the pathogenesis of various vascular diseases, such as atherosclerosis and restenosis. Among the mediators of VSMC during atherosclerosis development, platelet-derived growth factor (PDGF)-BB is a potent mitogen

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service