Skip to Content
Merck
All Photos(6)

Key Documents

SAB4504176

Sigma-Aldrich

Anti-phospho-ERK8 (pThr175+Tyr177) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

ERK7, MAPK15

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$529.00

$529.00


Check Cart for Availability


Select a Size

Change View
100 μG
$529.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$529.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 59 kDa

species reactivity

human, mouse, rat

concentration

~1 mg/mL

technique(s)

ELISA: 1:5000
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pThr175+pTyr177)

Gene Information

human ... MAPK15(225689)

General description

Mitogen-activated protein (MAP) kinase 15 (MAPK15) or extracellular signal-regulated kinase 8 (ERK8) belongs to the MAP kinase family. MAPK15 gene is located on human chromosome 8q24.3.

Immunogen

The antiserum was produced against synthesized peptide derived from human ERK8 around the phosphorylation site of Thr175 and Tyr177.

Immunogen Range: 141-190

Biochem/physiol Actions

Extracellular signal-regulated kinase 8 (ERK8)/mitogen-activated protein (MAP) kinase 15 aids in maintaining the integrity of the genome. Anaplastic thyroid carcinoma cells show high expression of MAPK15. It controls telomerase reverse transcriptase (hTERT) mRNA expression to regulate telomerase activity. It is highly essential to control the generation of reactive oxygen species. MAPK15 also plays a role in stabilizing proliferating cell nuclear antigen protein. MAPK15 might be involved in the progression of human cancer. Hence it might act as a candidate as an attractive target for cancer therapy.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dong-Hao Jin et al.
Oncotarget, 6(24), 20190-20203 (2015-06-03)
This study was aimed at understanding the functional and clinicopathological significance of MAPK15 alteration in gastric cancer. Genome-wide copy number alterations (CNAs) were first investigated in 40 gastric cancers using Agilent aCGH-244K or aCGH-400K, and copy number gains of MAPK15
David Colecchia et al.
The Journal of biological chemistry, 293(41), 15962-15976 (2018-08-23)
Autophagy, a pathway for bulk protein degradation and removal of damaged organelles, represents one of the major responses of cells to stress, thereby exerting a strict control on their correct functioning. Consequently, this process has been involved in the pathogenesis

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service