Skip to Content
Merck
All Photos(7)

Key Documents

SAB4504042

Sigma-Aldrich

Anti-phospho-Mst1/2 (pThr183) antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$622.00

$622.00


Check Cart for Availability


Select a Size

Change View
100 μG
$622.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$622.00


Check Cart for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 56 kDa

species reactivity

rat, mouse, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:40000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pThr183)

Gene Information

human ... MST1(4485)

Immunogen

The antiserum was produced against synthesized peptide derived from human Mst1/2 around the phosphorylation site of Thr183.

Immunogen Range: 149-198

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lezi Chen et al.
Cell death & disease, 10(4), 296-296 (2019-04-02)
Previous studies have revealed that dysregulation of long non-coding RNAs (lncRNAs) can facilitate carcinogenesis. This study aims to investigate the biological role of a certain lncRNA in cutaneous squamous cell carcinoma (CSCC). According to the data of TCGA database, high
Shanmin Zhao et al.
Cell death & disease, 11(1), 70-70 (2020-01-29)
Previous studies have shown that tumor necrosis factor (TNF)-α is a mediator of hepatotoxicity in liver injury. Moreover, TNF-α has also been reported to have a protective effect in liver regeneration, yet the function of TNF-α during liver injury remains
Xiehong Liu et al.
Journal of cellular physiology, 234(4), 5117-5133 (2018-09-27)
Heat-shock protein B1 (HSPB1) is a multifunctional protein that protects against oxidative stress; however, its function in antioxidant pathways remains largely unknown. Here, we sought to determine the roles of HSPB1 in H9c2 cells subjected to oxidative stress. Using nonreducing
Abdulhafiz Imam Aliagan et al.
Frontiers in endocrinology, 11, 579161-579161 (2020-11-17)
Introduction: Estrogen (17β-estradiol, E2) is well-known to induce cardioprotective effects against ischemia/reperfusion (I/R) injury. We recently reported that acute application of E2 at the onset of reperfusion in vivo induces cardioprotective effects against I/R injury via activation of its non-steroidal
Mengting Zhang et al.
Journal of neurochemistry, 143(2), 244-256 (2017-08-24)
The mammalian Ste20-like kinase 1 (Mst-1) is a serine-threonine kinase and a component of the Hippo tumor suppressor pathway, which reacts to pathologically relevant stress and regulates cell death. However, little is known about its role in spinal cord injury.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service