Skip to Content
Merck
All Photos(5)

Key Documents

SAB4503874

Sigma-Aldrich

Anti-phospho-GR (pSer226) antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$529.00

$529.00


Estimated to ship on26 April 2025



Select a Size

Change View
100 μG
$529.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$529.00


Estimated to ship on26 April 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 85 kDa

species reactivity

mouse, rat, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:5000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer226)

Gene Information

human ... GCR(2908)

Related Categories

General description

Glucocorticoid receptor (GR) gene/NR3C1 (nuclear receptor subfamily 3 group C member 1) is located on human chromosome 5q31. It belongs to the steroid receptor superfamily that has ligand-activated nuclear transcription factors.

Immunogen

The antiserum was produced against synthesized peptide derived from human GR around the phosphorylation site of Ser226.

Immunogen Range: 201-250

Biochem/physiol Actions

The glucocorticoid receptor (GR) gene/(NR3C1) is associated with asthma. It is required for glucose homeostasis and has been associated with hyperinsulinemia and increased fat deposition. GR also regulates cell differentiation, nervous and immune system functions.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Multiple promoters exist in the human GR gene, one of which is activated by glucocorticoids
Breslin MB, et al.
Molecular Endocrinology, 15(8), 1381-1395 (2001)
Glucocorticoid receptor polymorphism in obesity and glucose homeostasis
Majer-Lobodzinska A and Adamiec-Mroczek J
Advances in Clinical and Experimental Medicine : Official Organ Wroclaw Medical University, 26(1), 143-148 (2017)
Structure of the glucocorticoid receptor (NR3C1) gene 5' untranslated region: identification, and tissue distribution of multiple new human exon 1
Turner JD and Muller CP
Journal of Molecular Endocrinology, 35(2), 283-292 (2005)
Identification of polymorphisms in the human glucocorticoid receptor gene (NR3C1) in a multi-racial asthma case and control screening panel
Hawkins GA, et al.
Dna Sequence, 15(3), 167-173 (2004)
Alexander S Häusl et al.
Molecular psychiatry, 26(7), 3060-3076 (2021-03-03)
Disturbed activation or regulation of the stress response through the hypothalamic-pituitary-adrenal (HPA) axis is a fundamental component of multiple stress-related diseases, including psychiatric, metabolic, and immune disorders. The FK506 binding protein 51 (FKBP5) is a negative regulator of the glucocorticoid

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service