Skip to Content
Merck
All Photos(8)

Key Documents

SAB4503789

Sigma-Aldrich

Anti-phospho-Tyrosine Hydroxylase (pSer40) antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$529.00

$529.00


Check Cart for Availability


Select a Size

Change View
100 μG
$529.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$529.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 58 kDa

species reactivity

human, mouse, rat

concentration

~1 mg/mL

technique(s)

ELISA: 1:40000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer40)

Gene Information

human ... TH(7054)

General description

Tyrosine Hydroxylase (TH) is encoded by the gene mapped to human chromosome 11p15.5, close to the insulin (INS) and Harvey-ras-1 (HRAS) genes. The N-terminal end of the protein plays a vital role in maintaining intracellular stability of the enzyme. tyrosine hydroxylase is expressed in various tissues such as brain, adrenal medulla and sympathetically innervated tissues. The enzyme is characterized with a regulatory domain (R), a catalytic domain (C) at N-terminal end, and a coiled-coil domain at the C-terminal end.

Immunogen

The antiserum was produced against synthesized peptide derived from human Tyrosine Hydroxylase around the phosphorylation site of Ser40.

Immunogen Range: 41-90

Biochem/physiol Actions

Tyrosine Hydroxylase (TH) specifically catalyzes the hydroxylation of the amino acid L-tyrosine to 3, 4-dihydroxy-L-phenylalanine (L-DOPA). TH is a rate-limiting enzyme involved in the synthesis of catecholamine, involved in several brain functions, such as attention, memory, cognition, and emotion. Phosphorylation of the N-terminal portion of tyrosine hydroxylase controls the degradation of this enzyme by the ubiquitin-proteasome pathway. Alteration in the expression of the gene results in the pathogenesis of Parkinson′s disease (PD). Point mutation in the gene coding for TH leads to Segawa′s syndrome.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K L O'Malley et al.
Nucleic acids research, 16(10), 4437-4446 (1988-05-25)
The gene for the catecholamine biosynthetic enzyme, tyrosine hydroxylase (TH), has been previously mapped to human chromosome 11 p15.5 in the vicinity of the loci for insulin (INS) and for the oncogene Harvey Ras 1 (HRAS). Here we show that
B Lüdecke et al.
Human genetics, 95(1), 123-125 (1995-01-01)
We have examined the molecular basis of Segawa's syndrome in six families with seven affected children. In one family two siblings with this disease carried a point mutation in exon 11 of the tyrosine hydroxylase gene, resulting in an amino
Christopher Tolleson et al.
CNS & neurological disorders drug targets, 11(4), 381-386 (2012-04-10)
Tyrosine hydroxylase (TH) is the rate limiting step in the biosynthesis of dopamine and other catecholamines. Differences have been noted in concentration and availability of this enzyme and its cofactors in disease states such as Parkinson's disease (PD) which are
Akira Nakashima et al.
Biochemical and biophysical research communications, 407(2), 343-347 (2011-03-12)
Tyrosine hydroxylase (TH) is the rate-limiting enzyme in catecholamine biosynthesis, and its N-terminus plays a critical role in the intracellular stability of the enzyme. In the present study, we investigated the mechanism by which the N-terminal region of TH affects
S Colette Daubner et al.
Archives of biochemistry and biophysics, 508(1), 1-12 (2010-12-24)
Tyrosine hydroxylase is the rate-limiting enzyme of catecholamine biosynthesis; it uses tetrahydrobiopterin and molecular oxygen to convert tyrosine to DOPA. Its amino terminal 150 amino acids comprise a domain whose structure is involved in regulating the enzyme's activity. Modes of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service