Skip to Content
Merck
All Photos(2)

Key Documents

SAB4503786

Sigma-Aldrich

Anti-phospho-Trk B (pTyr705) antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$714.00

$714.00


Available to ship on23 April 2025Details



Select a Size

Change View
100 μG
$714.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$714.00


Available to ship on23 April 2025Details


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 91 kDa

species reactivity

mouse, rat, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:20000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pTyr705)

Gene Information

human ... NTRK2(4915)

General description

NTRK2 (neurotrophic tyrosine kinase receptor type 2) is also known as TRKB (tropomyosin receptor kinase B). It belongs to the Trk family of neurotrophin receptors. The NTRK2 gene is located on human chromosome 9q21.32.

Immunogen

The antiserum was produced against synthesized peptide derived from human Trk B around the phosphorylation site of Tyr705.

Immunogen Range: 671-720

Biochem/physiol Actions

In NB (neuroblastoma), NTRK2 (neurotrophic tyrosine kinase receptor type 2) controls the favorable and unfavorable clinical actions. It also controls the roles of glutamate receptor and synaptic plasticity in the amygdala.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A common NTRK2 variant is associated with emotional arousal and brain white-matter integrity in healthy young subjects
Spalek K, et al.
Translational Psychiatry (2016)
Entrectinib is a potent inhibitor of Trk-driven neuroblastomas in a xenograft mouse model
Iyer R, et al.
Cancer Letters, 372(2), 179-186 (2016)
Case Report: Next generation sequencing identifies a NAB2-STAT6 fusion in Glioblastoma
Diamandis P, et al.
Diagnostic Pathology (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service