Skip to Content
Merck
All Photos(5)

Key Documents

SAB4503756

Sigma-Aldrich

Anti-phospho-CaMK2 α/β/δ (pThr305) antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 54 kDa

species reactivity

rat, mouse, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:5000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pThr305)

Gene Information

General description

Calcium/calmodulin dependent protein kinase II β (CAMK2B) is part of the Ca2+/calmodulin-dependent protein kinase subfamily and the serine/threonine protein kinase family. The gene encoding CAMK2B is localized on human chromosome 7.

Immunogen

The antiserum was produced against synthesized peptide derived from human CaMK2 alpha/beta/delta around the phosphorylation site of Thr305.

Immunogen Range: 271-320

Biochem/physiol Actions

Calcium/calmodulin dependent protein kinase II β (CAMK2B) may bind to calmodulin. It has been associated with major depressive disorder.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Downregulation of ARFGEF1 and CAMK2B by promoter hypermethylation in breast cancer cells.
Kim JH
Bmb Reports, 44(8) null
Fine-mapping of vitiligo susceptibility loci on chromosomes 7 and 9 and interactions with NLRP1 (NALP1).
Jin Y
The Journal of Investigative Dermatology, 130(3), 774-783 (2010)
Xiang-Zi Zhang et al.
Bioscience reports, 39(12) (2019-11-27)
To observe changes in AMP-activated protein kinase (AMPK) activity and phosphorylation changes in AMPK signaling pathway in gastric smooth muscle cells of rats with diabetic gastroparesis (DGP), investigate the effect of AMPK on apoptosis and explore the underlying mechanism. After
Molecular cloning and sequence analyses of calcium/calmodulin-dependent protein kinase II from fetal and adult human brain. Sequence analyses of human brain calciuum/calmodulin-dependent protein kinase II.
Li G
Molecular Biology Reports, 28(1), 35-41 (2001)
Possible inhibitor from traditional Chinese medicine for the ? form of calcium-dependent protein kinase type II in the treatment of major depressive disorder.
Hung TC
BioMed Research International, 761849-761849 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service