Skip to Content
Merck
All Photos(1)

Key Documents

SAB4503736

Sigma-Aldrich

Anti-CDC25A antibody produced in rabbit

affinity isolated antibody

Synonym(s):

CDC25M3, Dual specificity phosphatase Cdc25A, M-phase inducer

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$589.00

$589.00


Check Cart for Availability


Select a Size

Change View
100 μG
$589.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

$589.00


Check Cart for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 59 kDa

species reactivity

human, mouse, rat

concentration

~1 mg/mL

technique(s)

ELISA: 1:10000
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CDC25A(993)

General description

Anti-CDC25A Antibody detects endogenous levels of total CDC25A protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human CDC25A.

Immunogen Range: 144-193

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jinhong Qi et al.
BMC cancer, 21(1), 819-819 (2021-07-17)
Cell division cycle 25A (CDC25A) is a well-recognized regulator of cell cycle progression and is involved in cancer development. This work focused on the function of CDC25A in cervical cancer cell growth and the molecules involved. A GEO dataset GSE63514

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service