Skip to Content
Merck
All Photos(1)

Key Documents

SAB4503685

Sigma-Aldrich

Anti-CEP135 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Centrosomal protein 4, Centrosomal protein of 135 kDa, Cep135

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$693.00

$693.00


Check Cart for Availability


Select a Size

Change View
100 μG
$693.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$693.00


Check Cart for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 133 kDa

species reactivity

mouse, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:40000
western blot: 1:500-1:1000

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CP135(9662)

General description

Anti-CEP135 Antibody detects endogenous levels of total CEP135 protein.
Cep135 (centrosomal protein 135) is a cartwheel protein. It has a coiled-coil domain and conserved regions near the N-terminus and the C-terminus, respectively. It is mapped to human chromosome 4q12.

Immunogen

The antiserum was produced against synthesized peptide derived from human CEP135.

Immunogen Range: 1081-1130

Application

Anti-CEP135, C-Terminal antibody has been used in immunofluorescence staining.

Biochem/physiol Actions

Mutation in Cep135 (centrosomal protein 135) result in primary microcephaly. It induces centriole assembly and stability. CEP135 helps to maintain the structure and organization of microtubules. This protein plays a major role in centriole biogenesis.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A truncating mutation of CEP135 causes primary microcephaly and disturbed centrosomal function
Hussain MS, et al.
American Journal of Human Genetics, 90(5), 871-878 (2012)
Control of apico-basal epithelial polarity by the microtubule minus-end-binding protein CAMSAP3 and spectraplakin ACF7
Noordstra I, et al.
Journal of Cell Science, 129(22), 4278-4288 (2016)
A Short CEP135 Splice Isoform Controls Centriole Duplication
Dahl KD, et al.
Current Biology, 25(19), 2591-2596 (2015)
A novel splice site mutation in CEP135 is associated with primary microcephaly in a Pakistani family
Farooq M, et al.
Journal of Human Genetics, 61(3), 271-273 (2016)
Fangrui Chen et al.
eLife, 11 (2022-07-06)
The major microtubule-organizing center (MTOC) in animal cells, the centrosome, comprises a pair of centrioles surrounded by pericentriolar material (PCM), which nucleates and anchors microtubules. Centrosome assembly depends on PCM binding to centrioles, PCM self-association and dynein-mediated PCM transport, but

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service