Skip to Content
Merck
All Photos(2)

Key Documents

SAB4503626

Sigma-Aldrich

Anti-EFTUD2 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

116 kDa, 116 kDa U5 small nuclear ribonucleoprotein component, KIAA0031, SNRP116, U5 snRNP- specific protein

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$520.00

$520.00


Check Cart for Availability


Select a Size

Change View
100 μG
$520.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$520.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 109 kDa

species reactivity

human, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:40000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... EFTUD2(9343)

General description

Anti-EFTUD2 Antibody detects endogenous levels of total EFTUD2 protein.
EFTUD2 (Elongation factor Tu GTP binding domain containing 2) is a member of the U5 small nuclear ribonucleoprotein particle (snRNP) located on chromosome region 17q21.31. It encodes a small GTPase (116kDa) component of the major spliceosome. It consists of a GTP-binding domain and several other conserved domains homologous to the translational elongation factor EF-2. Its N-terminal domain has an acidic domain.

Immunogen

The antiserum was produced against synthesized peptide derived from human EFTUD2.

Immunogen Range: 321-370

Application

Anti-EFTUD2 antibody produced in rabbit is suitable for immunohistochemistry and western blot.

Biochem/physiol Actions

EFTUD2 (Elongation factor Tu GTP binding domain containing 2) acts as a component of the spliceosome complex during translocation of mRNA on the ribosome. It produces mature mRNAs by binding to GTP during the processing of precursor mRNAs. Haploinsufficient genetic mutation in EFTUD2 causes a sporadic malformation syndrome, mandibulofacial dysostosis with microcephaly (MFDM), characterized with severe craniofacial abnormalities, microcephaly, growth delay, hearing loss, cleft palate, choanal atresia and dysmorphic features.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daphné Lehalle et al.
Human mutation, 35(4), 478-485 (2014-01-29)
Mandibulofacial dysostosis, Guion-Almeida type (MFDGA) is a recently delineated multiple congenital anomalies/mental retardation syndrome characterized by the association of mandibulofacial dysostosis (MFD) with external ear malformations, hearing loss, cleft palate, choanal atresia, microcephaly, intellectual disability, oesophageal atresia (OA), congenital heart
Margaret P Adam et al.
GeneReviews(?), 2014 Jul 03 (2014-07-08)
Mandibulofacial dysostosis with microcephaly (MFDM) is characterized by malar and mandibular hypoplasia; microcephaly (congenital or postnatal onset); malformations of the pinna, auditory canal, and/or middle ear (ossicles and semi-circular canals) with associated conductive hearing loss; and distinctive facial features (metopic
T Achsel et al.
Molecular and cellular biology, 18(11), 6756-6766 (1998-10-17)
The human small nuclear ribonucleoprotein (snRNP) U5 is biochemically the most complex of the snRNP particles, containing not only the Sm core proteins but also 10 particle-specific proteins. Several of these proteins have sequence motifs which suggest that they participate
S K Gandomi et al.
Clinical genetics, 87(1), 80-84 (2013-11-26)
Mandibulofacial dysostosis with microcephaly (MFDM) is a sporadic malformation syndrome with severe craniofacial abnormalities, microcephaly, developmental delay, and dysmorphic features. Most cases of clinically diagnosed MFDM remain genetically unexplained, and to the best of our knowledge a total of 35

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service