Skip to Content
Merck
All Photos(3)

Key Documents

SAB4503514

Sigma-Aldrich

Anti-Cyclin E1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

CCNE, CCNE1, G1/S-specific cyclin E1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$589.00

$589.00


Check Cart for Availability


Select a Size

Change View
100 μG
$589.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$589.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 47 kDa

species reactivity

human

concentration

~1 mg/mL

technique(s)

ELISA: 1:10000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CCNE1(898)

General description

Anti-Cyclin E1 Antibody detects endogenous levels of total Cyclin E1 protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human Cyclin E1.

Immunogen Range: 361-410

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gerwin Heller et al.
The Journal of pathology, 245(4), 387-398 (2018-03-24)
Deregulated DNA methylation leading to transcriptional inactivation of certain genes occurs frequently in non-small-cell lung cancers (NSCLCs). As well as protein-coding genes, microRNA (miRNA)-coding genes may be targets for methylation in NSCLCs; however, the number of known methylated miRNA genes
Cheng Zhang et al.
Oncology letters, 13(4), 2059-2070 (2017-04-30)
Human bone marrow stromal cells (hBMSCs) may contribute to the growth of tyrosine kinase inhibitor (TKI)-resistant chronic myelogenous leukemia (CML). However, there are certain differences in biology between CML and Philadelphia-positive acute lymphoblastic leukemia (Ph+ ALL). Little is known about

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service