Skip to Content
Merck
All Photos(1)

Key Documents

SAB4503385

Sigma-Aldrich

Anti-USP30 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Deubiquitinating enzyme 30, USP30, Ubiquitin carboxyl-terminal hydrolase 30, Ubiquitin thioesterase 30, Ubiquitin-specific-processing protease 30

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$693.00

$693.00


Check Cart for Availability


Select a Size

Change View
100 μG
$693.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$693.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 58 kDa

species reactivity

human, rat, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:10000
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... USP30(84749)

General description

Anti-USP30 Antibody detects endogenous levels of total USP30 protein.
USP30 (ubiquitin specific peptidase 30) is ubiquitin-specific deubiquitylase. It is located in the mitochondria. It is mapped to human chromosome 12q24.

Immunogen

The antiserum was produced against synthesized peptide derived from human USP30.

Immunogen Range: 31-80

Application

Anti-USP30, N-Terminal antibody has been used in western blotting.

Biochem/physiol Actions

USP30 controls atypical ubiquitin chains on mitochondria and PARK2 (parkinson juvenile disease protein 2) mediated mitophagy.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

USP30 and parkin homeostatically regulate atypical ubiquitin chains on mitochondria
Cunningham CN, et al.
Nature Cell Biology, 17(2), 160-160 (2015)
A genome-wide study reveals rare CNVs exclusive to extreme phenotypes of Alzheimer disease
Rovelet-Lecrux A, et al.
European Journal of Human Genetics, 20(6), 613-617 (2012)
Deubiquitinating enzymes regulate PARK2-mediated mitophagy
Wang Y, et al.
Autophagy, 11(4), 595-606 (2015)
Daily heat stress treatment rescues denervation-activated mitochondrial clearance and atrophy in skeletal muscle
Tamura Y, et al.
The Journal of Physiology, 593(12), 2707-2720 (2015)
Ding Yan et al.
Cell death discovery, 7(1), 211-211 (2021-08-13)
Deubiquitinates (DUBs) have been suggested as novel promising targets for cancer therapies. Accumulating experimental evidence suggests that some metal compounds have the potential to induce cancer cell death via inhibition of DUBs. We previously reported that auranofin, a gold(I)-containing agent

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service