Skip to Content
Merck
All Photos(2)

Key Documents

SAB4503371

Sigma-Aldrich

Anti-NT5C1A antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Cytosolic 5′-nucleotidase 1A, Cytosolic 5′-nucleotidase IA, cN-I, cN-IA, cN1A

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$693.00

$693.00


Check Cart for Availability


Select a Size

Change View
100 μG
$693.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$693.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 41 kDa

species reactivity

mouse, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:20000
immunofluorescence: 1:100-1:500
western blot: 1:500-1:1000

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... 5NT1A(84618)

General description

Anti-NT5C1A Antibody detects endogenous levels of total NT5C1A protein. The NT5C1A (5′-nucleotidase, cytosolic IA) gene is mapped to human chromosome 1p34. The protein encoded is ubiquitously expressed and is localized to the cytoplasm.

Immunogen

The antiserum was produced against synthesized peptide derived from human NT5C1A.

Immunogen Range: 151-200

Biochem/physiol Actions

NT5C1A (5′-nucleotidase, cytosolic IA) forms a tetramer with AMP (adenosine monophosphate) and serves as a primary substrate in adenosine pathway to control the level of metabolite produced. NT5C1A catalyzes the dephosphorylation of AMP to adenosine.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pharmacogenetic characterization of naturally occurring germline NT5C1A variants to chemotherapeutic nucleoside analogs.
Saliba J, et al.
Pharmacogenetics and Genomics, 26(6), 271-271 (2016)
Expression of cytosolic 5? nucleotidase does not correlate with expression of oxidative metabolism marker: myoglobine in human skeletal muscles.
Lechward K and Tkacz-Stachowska K
Acta biochimica et biophysica Sinica, 41(4), 280-284 (2009)
Suppression of 5'nucleotidase enzymes promotes AMPK phosphorylation and metabolism in human and mouse skeletal muscle.
Kulkarni S S, et al.
The Journal of Biological Chemistry (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service