Skip to Content
Merck
All Photos(1)

Key Documents

SAB4503210

Sigma-Aldrich

Anti-FGF23 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

FGF-23, Fibroblast growth factor 23, HYPF, Tumor-derived hypophosphatemia-inducing factor

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$589.00

$589.00


Check Cart for Availability


Select a Size

Change View
100 μG
$589.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$589.00


Check Cart for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 27 kDa

species reactivity

rat, human, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:20000
immunofluorescence: 1:100-1:500

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FGF23(8074)

General description

Fibroblast growth factor-23 (FGF-23) is a bone-derived hormone which is secreted by osteocytes and osteoblasts. It is part of the atypical FGF subfamily. The gene encoding it is localized on human chromosome 12p13. Anti-FGF23 Antibody detects endogenous levels of total FGF23 protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human FGF23.

Immunogen Range: 151-200

Biochem/physiol Actions

The FGF family plays a central role during prenatal development and postnatal growth and regeneration of a variety of tissues, by promoting cellular proliferation and differentiation. Fibroblast growth factor-23, -21 and -19 (FGF-23, FGF-21 and FGF-19) act as circulating hormones and require the participation of a Klotho protein as a co-receptor for their signaling. The signaling receptor for FGF-23, a Klotho-FGFR1 (IIIc) complex, is an essential regulator of the renal sodium phosphate co-transporter and key vitamin D-metabolizing enzymes cytochrome P450 family 27 subfamily B member 1 (CYP27B1) and cytochrome P450 family 24 subfamily A member 1 (CYP24A1). FGF-23 acts in the kidney to regulate phosphate homeostasis and vitamin D metabolism.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Klotho converts canonical FGF receptor into a specific receptor for FGF23.
Urakawa I
Nature, 444(7120), 770-774 (2006)
Interplay between vitamin D and the drug metabolizing enzyme CYP3A4.
Wang Z
The Journal of Steroid Biochemistry and Molecular Biology, 136, 54-58 (2013)
Fibroblast growth factor 23 and bone mineralisation.
Guo YC and Yuan Q
International Journal of Oral Science, 7(1), 8-13 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service