Skip to Content
Merck
All Photos(2)

Key Documents

SAB4503170

Sigma-Aldrich

Anti-FZD3 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

FZD3, Frizzled-3, Fz-3, hFz3

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 76 kDa

species reactivity

mouse, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:1000
immunofluorescence: 1:100-1:500
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FZD3(7976)

Related Categories

General description

Anti-FZD3 Antibody detects endogenous levels of total FZD3 protein.
The FZD3 (frizzled class receptor 3) gene is mapped to human chromosome 8p21.1. It codes for a receptor with seven-transmembrane domain and is expressed in human melanocytes.

Immunogen

The antiserum was produced against synthesized peptide derived from human FZD3.

Immunogen Range: 141-190

Application

Anti-FZD3 antibody produced in rabbit has been used in immunohistochemistry.

Biochem/physiol Actions

Mutation in FZD3 (frizzled class receptor 3) gene might account for Hirschsprung disease, a birth defect lacking the intrinsic ganglion cells of the lower intestine. FZD3 regulates the growth of longitudinal axon tracts in the central nervous system. FZD3 mediates the dynamics of axon within the enteric, sympathetic and peripheral nervous systems. FZD3 regulates planar cell polarity. Abnomal FZD3 gene methylation causes chromatin structure modifications, associated with congenital hydrocephalus.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jie Xie et al.
International journal of developmental neuroscience : the official journal of the International Society for Developmental Neuroscience, 66, 24-32 (2017-12-03)
Previous studies have demonstrated that both Wnt5a and Sonic hedgehog (Shh) are involved in regulating the pathfinding of descending serotonergic (5-HT, 5-hydroxytryptamine) axons in an opposite manner in the brainstem. Shh and Wnt signaling pathways interact to guide post-crossing commissural
The roles of Frizzled-3 and Wnt3a on melanocyte development: in vitro studies on neural crest cells and melanocyte precursor cell lines.
Chang CH, et al.
Journal of Dermatological Science, 75(2), 100-108 (2014)
Impaired methylation modifications of FZD3 alter chromatin accessibility and are involved in congenital hydrocephalus pathogenesis.
Wang L, et al.
Brain Research, 1569, 48-56 (2014)
Ateeque Siddique et al.
Canadian journal of physiology and pharmacology, 96(2), 208-214 (2017-12-16)
The cellular mechanisms that induce calcific aortic stenosis are yet to be unraveled. Wnt signaling is increasingly being considered as a major player in the disease process. However, the presence of Wnt Frizzled (Fzd) receptors and co-receptors LRP5 and 6
Genotype?Phenotype Association Studies of Chromosome 8p Inverted Duplication Deletion Syndrome
Fisch GS, et al.
Behavior Genetics, 41(3), 373-380 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service