Skip to Content

We are planning system maintenance on January 25, 2025, from 1:00 PM to 4:00 PM CST. This will impact both web and offline transactions, including online orders, quotes, price and availability checks, and order status inquiries. We apologize for any inconvenience.

Merck
All Photos(2)

Key Documents

SAB4502870

Sigma-Aldrich

Anti-STAT2 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

STAT2, Signal transducer and activator of transcription 2, p113

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
USD 585.00
5 G
USD 2,340.00

Vendor SKU: BB-0496-1G

USD 585.00


Ships in 7 days - from Aldrich Partner.


Select a Size

Change View
1 G
USD 585.00
5 G
USD 2,340.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

conjugate:
unconjugated
application:
ELISA
IHC
WB
clone:
polyclonal
species reactivity:
human, rat
citations:
technique(s):
ELISA: 1:5000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

Vendor SKU: BB-0496-1G

USD 585.00


Ships in 7 days - from Aldrich Partner.

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 97 kDa

species reactivity

human, rat

concentration

~1 mg/mL

technique(s)

ELISA: 1:5000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
1250591.250631.25062
ligand

(Trimethylammoniumethyl)

ligand

diethyl-(2-hydroxypropyl)aminoethyl

ligand

iminodiacetic acid

ligand

(Carboxyethyl )

capacity

~100 mg, BSA binding capacity

capacity

~100 mg, BSA binding capacity

capacity

~60 mL, resin binding capacity

capacity

~100 mL, resin binding capacity

form

prepacked column

form

prepacked column

form

prepacked column

form

prepacked column

material

polypropylene housing

material

polypropylene housing

material

polypropylene housing

material

polypropylene housing

application(s)

gene therapy
recombinant protein
vaccine development

application(s)

gene therapy
recombinant protein
vaccine development

application(s)

recombinant protein

application(s)

mAb
recombinant protein
vaccine development

General description

Anti-STAT2 Antibody detects endogenous levels of total STAT2 protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human STAT2.

Immunogen Range: 656-705

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service