Skip to Content
Merck
All Photos(2)

Key Documents

SAB4502671

Sigma-Aldrich

Anti-RPL39, N-Terminal antibody produced in rabbit

affinity isolated antibody

Synonym(s):

60S ribosomal protein L39

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$693.00

$693.00


Check Cart for Availability


Select a Size

Change View
100 μG
$693.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$693.00


Check Cart for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 6 kDa

species reactivity

human, rat, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:60000
immunofluorescence: 1:100-1:500
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RPL39(6170)

General description

Ribosomal protein L39 (RPL39), a component of the 60S ribosomal complex. The RPL39 gene is mapped on human chromosome XQ24.

Immunogen

The antiserum was produced against synthesized peptide derived from human RPL39.

Immunogen Range: 1-50

Biochem/physiol Actions

Ribosomal protein L39 (RPL39) plays a major role in spermatogenesis and translation. It might participate in ribosome biogenesis. Mutations in the RPL39 gene are associated with breast cancer and lung metastasis. RPL39 is capable of disrupting nitric oxide synthase and hypoxia signaling pathway. Overexpression of RPL39 can enhance wound-healing in MDAMB231 and BT549 cells. Antibody detects endogenous levels of total RPL39 protein.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bhuvanesh Dave et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(24), 8838-8843 (2014-05-31)
We previously described a gene signature for breast cancer stem cells (BCSCs) derived from patient biopsies. Selective shRNA knockdown identified ribosomal protein L39 (RPL39) and myeloid leukemia factor 2 (MLF2) as the top candidates that affect BCSC self-renewal. Knockdown of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service