Skip to Content
Merck
All Photos(1)

Key Documents

SAB4502482

Sigma-Aldrich

Anti-PTGER3 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

EP3 subtype, PGE receptor, PGE2-R, Prostaglandin E2 receptor EP3 subtype, Prostanoid EP3 receptor

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$589.00

$589.00


Check Cart for Availability


Select a Size

Change View
100 μG
$589.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$589.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 43 kDa

species reactivity

human

concentration

~1 mg/mL

technique(s)

ELISA: 1:10000
immunofluorescence: 1:100-1:500

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PTGER3(5733)

Related Categories

General description

The PTGER3 (prostaglandin E receptor 3) gene is mapped to human chromosome 1p31.1 and encodes a Gαi coupled receptor protein. It is predominantly localized to the brain in the neurons and activated glial cells. Anti-PTGER3 antibody detects endogenous levels of total PTGER3 protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human PTGER3.

Immunogen Range: 16-65

Biochem/physiol Actions

Prostaglandin E receptor 3 (PTGER3) serves as a receptor for prostaglandin (PG) E1 and E2, to carry out a number of physiological processes. PTGER3 specifically functions to reduce adenylyl cyclase action. It is involved in the activation of the Ras signaling pathway.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inhibition of prostaglandin E2 receptor EP3 mitigates thrombin-induced brain injury
Han, X, et al.
Journal of Cerebral Blood Flow and Metabolism, 36(6), 1059-1074 (2016)
Tuberin Regulates Prostaglandin Receptor-Mediated Viability, via Rheb, in mTORC1-Hyperactive Cells
Li C, et al.
Molecular Cancer Research (2017)
The key residue within the second extracellular loop of human EP3 involved in selectively turning down PGE 2-and retaining PGE 1-mediated signaling in live cells
Akasaka H
Archives of Biochemistry and Biophysics, 616, 20-29 (2017)
Loss of heterozygosity at chromosome 1p in different solid human tumours: association with survival.
Ragnarsson G
British Journal of Cancer, 79(9-10), 1468-1474 (1999)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service