Skip to Content
Merck
All Photos(2)

Key Documents

SAB4502419

Sigma-Aldrich

Anti-Peripherin antibody produced in rabbit

affinity isolated antibody

Synonym(s):

PERI, PRPH

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$589.00

$589.00


Check Cart for Availability


Select a Size

Change View
100 μG
$589.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$589.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 53 kDa

species reactivity

rat, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:5000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PRPH(5630)

General description

Anti-Peripherin antibody detects endogenous levels of total Peripherin protein.
The PRPH (peripherin) gene is mapped to human chromosome 12q13.12. This gene codes for a 57 kDa intermediate filament protein. Peripherin is predominantly expressed in the peripheral neurons.

Immunogen

The antiserum was produced against synthesized peptide derived from human Peripherin.

Immunogen Range: 421-470

Biochem/physiol Actions

Peripherin is known to be associated with the mechanism of organelle transport, function and positioning. It also forms an important part of the cytoskeleton. Mutation in PRPH (peripherin) is observed in amyotrophic lateral sclerosis. Peripherin participates in the elongation of motor axons. Upregulation of PRPH is noticed in neuronal injury.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joshua J Emrick et al.
Proceedings of the National Academy of Sciences of the United States of America, 115(51), E12091-E12100 (2018-11-23)
Atopic dermatitis (AD) is the most common skin disease in children. It is characterized by relapsing inflammation, skin-barrier defects, and intractable itch. However, the pathophysiology of itch in AD remains enigmatic. Here, we examine the contribution of Tmem79, an orphan
Paulo Victor Sgobbi de Souza et al.
Arquivos de neuro-psiquiatria, 73(12), 1026-1037 (2015-10-16)
Amyotrophic lateral sclerosis represents the most common neurodegenerative disease leading to upper and lower motor neuron compromise. Although the vast majority of cases are sporadic, substantial gain has been observed in the knowledge of the genetic forms of the disease
Charcot-Marie-Tooth type 2B disease-causing RAB7A mutant proteins show altered interaction with the neuronal intermediate filament peripherin.
Cogli L
Acta Neuropathologica, 125(2), 257-272 (2013)
Jesse McLean et al.
Journal of neurochemistry, 114(4), 1177-1192 (2010-06-11)
Peripherin is a type III intermediate filament protein that is up-regulated during neuronal injury and is a major component of pathological inclusions found within degenerating motor neurons of patients with amyotrophic lateral sclerosis (ALS). The relationship between these inclusions and
Stephanie Shiers et al.
Pain, 161(11), 2494-2501 (2020-08-23)
SARS-CoV-2 has created a global crisis. COVID-19, the disease caused by the virus, is characterized by pneumonia, respiratory distress, and hypercoagulation and can be fatal. An early sign of infection is loss of smell, taste, and chemesthesis-loss of chemical sensation.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service