Skip to Content
Merck
All Photos(2)

Documents

SAB4502029

Sigma-Aldrich

Anti-NPY2R antibody produced in rabbit

affinity isolated antibody

Synonym(s):

NPY-Y2 receptor, NPY2-R, NPY2R, Neuropeptide Y receptor type 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 42 kDa

species reactivity

human, mouse, rat

concentration

~1 mg/mL

technique(s)

ELISA: 1:1000
immunofluorescence: 1:100-1:500
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NPY2R(4887)

Related Categories

General description

The NPY2R (neuropeptide Y receptor Y2) gene is mapped to human chromosome 4q32.1 and codes for a G-protein-coupled receptor. NPY2R is predominantly expressed in central nervous system including hypothalamus, brain stem, hippocampus, lateral septum and amygdala. NPY2 receptors are also distributed in the liver, spleen, intestine, adipose tissue and muscle.

Immunogen

The antiserum was produced against synthesized peptide derived from human NPY2R.

Immunogen Range: 1-50

Biochem/physiol Actions

NPY2R (neuropeptide Y receptor Y2) acts as a presynaptic autoreceptor and heteroreceptor. It regulates the release of both endogenous NPY and neurotransmitters such as glutamate and γ–aminobutyric acid (GABA). NPY exerts its anxiolytic function via the NPY2R. Deletion of NPY2R results in retardation of the anticonvulsant actions of NPY. NPY2R plays an important role in the process of bone formation.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Debanjan Chakroborty et al.
British journal of cancer, 127(8), 1440-1449 (2022-07-29)
Resistance to anti-angiogenic therapies targeting vascular endothelial growth factor-A (VEGF-A) stems from VEGF-A independent angiogenesis mediated by other proangiogenic factors. Therefore identifying these factors in colon adenocarcinoma (CA) will reveal new therapeutic targets. Neuropeptide Y (NPY) and Y2 receptor (Y2R)
Tao Yang et al.
Cellular & molecular biology letters, 28(1), 60-60 (2023-07-28)
Sebaceous glands (SGs) synthesize and secret sebum to protect and moisturize the dermal system via the complicated endocrine modulation. Dysfunction of SG are usually implicated in a number of dermal and inflammatory diseases. However, the molecular mechanism behind the differentiation
Unwinding of the C-Terminal Residues of Neuropeptide Y is critical for Y2 Receptor Binding and Activation.
Kaiser A
Angewandte Chemie (International Edition in English), 54(25), 7446-7449 (2015)
Mayur J Patil et al.
PloS one, 13(6), e0198601-e0198601 (2018-06-05)
Peptidergic sensory neurons play a critical role in nociceptive pathways. To precisely define the function and plasticity of sensory neurons in detail, new tools such as transgenic mouse models are needed. We employed electrophysiology and immunohistochemistry to characterize in detail
Viswanthram Palanivel et al.
Cells, 11(22) (2022-11-27)
Neuropeptide Y (NPY), a sympathetic neurotransmitter, is involved in various physiological functions, and its dysregulation is implicated in several neurodegenerative diseases. Glutamate excitotoxicity, endoplasmic reticulum (ER) stress, and oxidative stress are the common mechanisms associated with numerous neurodegenerative illnesses. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service