Skip to Content
Merck
All Photos(1)

Key Documents

SAB4501145

Sigma-Aldrich

Anti-LATS1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Kinase phosphorylated during mitosis protein, Large tumor suppressor homolog 1, Serine/threonine-protein kinase kpm, WARTS protein kinase, h-warts/ Large tumor suppressor homolog 2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$589.00

$589.00


Estimated to ship on29 May 2025



Select a Size

Change View
100 μG
$589.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$589.00


Estimated to ship on29 May 2025


biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 126 kDa

species reactivity

human, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:40000
immunohistochemistry: 1:50-1:100

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LATS1(9113)

General description

Anti-LATS1 Antibody detects endogenous levels of total LATS1 protein.
The LATS1 (large tumor suppressor kinase 1) gene encodes a serine/threonine kinase and is located on human chromosome 6q24-25. It is present in the mitotic apparatus. It is also called as WARTS.

Immunogen

The antiserum was produced against synthesized peptide derived from human LATS1/2.

Immunogen Range: 1041-1090

Biochem/physiol Actions

The LATS1 (large tumor suppressor kinase 1) protein repress the multiplication and invasion of cervical cancer. It acts as a cell cycle inhibitor. Low level expression of LATS1 is associated with the progression and prognosis of glioma. It is a tumor suppressor gene, thereby it participates in various mitotic processes.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

LATS1 suppresses proliferation and invasion of cervical cancer
Deng J, et al.
Molecular Medicine Reports, 15(4), 1654-1660 (2017)
Underexpression of LATS1 TSG in colorectal cancer is associated with promoter hypermethylation
Wierzbicki PM, et al.
World Journal of Gastroenterology, 19(27), 4363-4373 (2013)
Decreased expression of LATS1 is correlated with the progression and prognosis of glioma
Ji T, et al.
Journal of Experimental & Clinical Cancer Research (2012)
Di Li et al.
CNS neuroscience & therapeutics, 24(10), 906-916 (2018-03-01)
To investigate the roles of Lats1/p-YAP1 pathway in TBI-induced neuronal apoptosis and neurological deficits in rats. We found that Lats1 and YAP1 were expressed in cerebral cortex neurons of Sprague-Dawley rats, and the phosphorylation levels of Lats1 and YAP1 in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service