Skip to Content
Merck
All Photos(1)

Key Documents

SAB4500930

Sigma-Aldrich

Anti-MAST3 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

KIAA0561, MAST3, Microtubule-associated serine/threonine-protein kinase 3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$520.00

$520.00


Check Cart for Availability


Select a Size

Change View
100 μG
$520.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$520.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 143 kDa

species reactivity

mouse, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:10000
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MAST3(23031)

General description

Anti-MAST3 Antibody detects endogenous levels of total MAST3 protein. Microtubule associated serine/threonine kinase 3 (MAST3) belongs to the MAST kinase family. It is composed of 1309 amino acids. The MAST3 gene is 53.9kb long and has 27 exons. It is localized on human chromosome 19p13.11. MAST3 possesses four α-helix bundles having various domains, including a protein kinase domain and a PDZ domain.

Immunogen

The antiserum was produced against synthesized peptide derived from human MAST3.

Immunogen Range: 1031-1080

Biochem/physiol Actions

Microtubule associated serine/threonine kinase 3 (MAST3) modulates the activity of nuclear factor κ-B (NF-κB). It may have a role in inflammatory pathways. Knockdown of MAST3 has been shown to reduce NF-κB activity, which is dependent on toll-like receptor-4.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Genomic alterations identified by array comparative genomic hybridization as prognostic markers in tamoxifen-treated estrogen receptor-positive breast cancer
Wonshik Han
BMC Cancer (2006)
MAST3: a novel IBD risk factor that modulates TLR4 signaling.
Labbe C
Genes and Immunity (2008)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service