Skip to Content
Merck
All Photos(2)

Key Documents

SAB4500395

Sigma-Aldrich

Anti-Collagen XII α1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

COCA1, Collagen α-1(XII) chain

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$589.00

$589.00


Check Cart for Availability


Select a Size

Change View
100 μG
$589.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$589.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 333 kDa

species reactivity

human

concentration

~1 mg/mL

technique(s)

ELISA: 1:1000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... COL12A1(1303)

General description

Anti-Collagen XII α1 Antibody detects endogenous levels of total Collagen XII α1 protein.
Collagen XII α1 belongs to the FACIT collagens (fibril-associated collagens with interrupted triple helices) family. It codes for the alpha chain of type XII collagen. It is located on human chromosome 6q13.

Immunogen

The antiserum was produced against synthesized peptide derived from human Collagen XII alpha1.

Immunogen Range: 1481-1530

Application

Anti-Collagen XII α1 antibody has been used in immunohistochemistry.[1]

Biochem/physiol Actions

Collagen XII α1 participates in a benign bone tumor, subungual exostosis. Collagen XII organize the collagen fibrils and thereby preserves the bone and muscle integrity. Mutations in collagen XII results in Ehlers–Danlos/myopathy overlap syndrome.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Collagen XII: protecting bone and muscle integrity by organizing collagen fibrils.
Chiquet M, et al.
The International Journal of Biochemistry & Cell Biology, 53, 51-54 (2014)
Mutations in the collagen XII gene define a new form of extracellular matrix-related myopathy.
Hicks D, et al.
Human Molecular Genetics, 23(9), 2353-2363 (2013)
Ivana Zvackova et al.
Frontiers in physiology, 8, 554-554 (2017-08-22)
Development of mammalian teeth and surrounding tissues includes time-space changes in the extracellular matrix composition and organization. This requires complex control mechanisms to regulate its synthesis and remodeling. Fibril-associated collagens with interrupted triple helices (FACITs) and a group of small
Taketoshi Yasuda et al.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc, 22(11), 1499-1506 (2009-08-04)
Chondromyxoid fibroma, a rare benign bone tumor, may be mistaken for chondrosarcoma. Although cytogenetic studies of chondromyxoid fibroma are few, rearrangements of the long arm of chromosome 6, frequently expressed as an inv(6)(p25q13), are prominent. In this study, conventional cytogenetic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service