Skip to Content
Merck
All Photos(2)

Key Documents

SAB4500001

Sigma-Aldrich

Anti-Cadherin-pan antibody produced in rabbit

affinity isolated antibody

Synonym(s):

CAM 120/80, CD324, Cadherin-1, Epithelial cadherin, Uvomorulin

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$589.00

$589.00


Check Cart for Availability


Select a Size

Change View
100 μG
$589.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$589.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 97 kDa

species reactivity

human, rat, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:20000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CDH1(999)

General description

Anti-Cadherin-pan Antibody detects endogenous levels of total Cadherin-pan protein.
CDH1 (cadherin 1) codes for a transmembrane glycoprotein E-cadherin that is located on human chromosome 16q22.1. It is a tumor suppressor gene, which is also known as Cdh1p/Hct1p, Fizzy-related, Ste9, or Srw1.

Immunogen

The antiserum was produced against synthesized peptide derived from human Cadherin.

Immunogen Range: 833-882

Biochem/physiol Actions

CDH1 (cadherin 1) helps to conserve the homophilic cell-cell adhesion in epithelial tissues. Inadequate expression of CDH1 results in colon cancer and renal cell carcinoma (RCC).

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

CDH1 promoter hypermethylation and E-cadherin protein expression in infiltrating breast cancer
Caldeira JR, et al.
BMC Cancer, 6, 48-48 (2006)
Mitotic regulation of the APC activator proteins CDC20 and CDH1
Kramer ER, et al.
Molecular Biology of the Cell, 11(5), 1555-1569 (2000)
Identification of downstream metastasis-associated target genes regulated by LSD1 in colon cancer cells
Chen J, et al.
Oncotarget, 8(12), 19609-19630 (2017)
Reduced E-cadherin facilitates renal cell carcinoma progression by WNT/?-catenin signaling activation
Zhang X, et al.
Oncotarget, 8(12), 19566-19576 (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service