Skip to Content
Merck
All Photos(1)

Key Documents

SAB4301794

Sigma-Aldrich

Anti-SLC4A3

affinity isolated antibody

Synonym(s):

AE3, SLC2C

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$532.00

$532.00


Check Cart for Availability


Select a Size

Change View
100 μL
$532.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$532.00


Check Cart for Availability

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

human, rat, mouse

technique(s)

western blot: 1:200-1:1000

accession no.

NP_005061

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SLC4A3 (6508)

Immunogen

Synthetic peptide of human SLC4A3

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in pH7.4 PBS, 0.05% NaN3, 40% Glycerol.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service