Skip to Content
Merck
All Photos(2)

Key Documents

SAB4301759

Sigma-Aldrich

Anti-PRDX4

affinity isolated antibody

Synonym(s):

AOE37-2, AOE372, HEL-S-97n, PRX-4

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$532.00

$532.00


Check Cart for Availability


Select a Size

Change View
100 μL
$532.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$532.00


Check Cart for Availability

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

mouse, human, rat

technique(s)

immunohistochemistry: 1:20-1:100
western blot: 1:200-1:1000

accession no.

NP_006397

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PRDX4(10549)

Immunogen

Synthetic peptide of human PRDX4

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in pH7.4 PBS, 0.05% NaN3, 40% Glycerol.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Blendi Ura et al.
International journal of molecular medicine, 44(6), 2329-2335 (2019-10-23)
Uterine leiomyomas are benign smooth muscle cell tumors originating from the myometrium. The present study focused on leiomyoma and myometrium phosphoproteome enrichment by using immobilized metal affinity chromatography (IMAC). The phosphoproteome was analyzed by two‑dimensional gel electrophoresis coupled with mass
Nicole M Sayles et al.
Cell reports, 38(10), 110475-110475 (2022-03-10)
Mitochondrial cardiomyopathies are fatal diseases, with no effective treatment. Alterations of heart mitochondrial function activate the mitochondrial integrated stress response (ISRmt), a transcriptional program affecting cell metabolism, mitochondrial biogenesis, and proteostasis. In humans, mutations in CHCHD10, a mitochondrial protein with

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service