Skip to Content
Merck
All Photos(2)

Key Documents

SAB4301756

Sigma-Aldrich

Anti-PF4

affinity isolated antibody

Synonym(s):

CXCL4, PF-4, SCYB4

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$532.00

$532.00


Check Cart for Availability


Select a Size

Change View
100 μL
$532.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

$532.00


Check Cart for Availability

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:20-1:100

accession no.

NP_002610

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PF4(5196)

General description

Platelet factor-4 (PF-4) is a CXC chemokine that is expressed in megakaryocytes and stored in the alpha-granules of platelets. The gene encoding this protein is localized on human chromosome 4.

Immunogen

Synthetic peptide of human PF4

Biochem/physiol Actions

Platelet factor-4 (PF-4) is chemotactic towards neutrophils and monocytes and has been shown to inhibit angiogenesis. This protein associates with heparin and neutralizes it. It also neutralizes various negatively charged proteoglycans.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in pH7.4 PBS, 0.05% NaN3, 40% Glycerol.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

CXCL4 Plasma Levels Are Not Associated with the Extent of Coronary Artery Disease or with Coronary Plaque Morphology.
Erbel C
PLoS ONE, 10(11), ee0141693-ee0141693 (2015)
Chemokines and Chemokine Receptors
Manel Juan
Wiley Interdisciplinary Reviews. RNA null
CXCL4-platelet factor 4, heparin-induced thrombocytopenia and cancer.
Sandset PM
Thrombosis Research, 129, S97-100 (2012)
Transformation-associated cytokine 9E3/CEF4 is chemotactic for chicken peripheral blood mononuclear cells.
Barker KA
Journal of Virology, 67(6), 3528-3533 (1993)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service