Skip to Content
Merck
All Photos(1)

Key Documents

SAB4301413

Sigma-Aldrich

Anti-phospho-ATF1 (pSer63) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

ATF1, Activating transcription factor 1, Cyclic-AMP-dependent transcription factor ATF-1, TCR-ATF1, TREB36 protein

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$463.00

$463.00


Estimated to ship on13 May 2025



Select a Size

Change View
100 μL
$463.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$463.00


Estimated to ship on13 May 2025


biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

29 kDa

species reactivity

mouse, human

concentration

1.0 mg/mL

technique(s)

western blot: 1:500-1:1000 (Cell Lysate)

isotype

IgG

accession no.

NP_005162.1

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer63)

Gene Information

human ... ATF1(466)

Specificity

The antibody detects endogenous level of ATF1 only when phosphorylated at serine 63.

Immunogen

Peptide sequence around phosphorylation site of serine 63(R-P-S(p)-Y-R) derived from Human ATF1 .

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service