Skip to Content
Merck
All Photos(2)

Key Documents

SAB4301002

Sigma-Aldrich

Anti-TAB1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

3′-Tab1, MAP3K7IP1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$470.00

$470.00


Check Cart for Availability


Select a Size

Change View
100 μL
$470.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$470.00


Check Cart for Availability

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

55 kDa

species reactivity

human

concentration

2.1 mg/mL

technique(s)

immunohistochemistry: 1:100- 1:300
western blot: 1:200-1:1000 (Cell Lysate)

isotype

IgG

accession no.

NP_006107.1

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TAB1(10454)

Specificity

The antibody detects endogenous levels of total TAB1 protein.

Immunogen

Fusion protein corresponding to a region derived from internal residues of human TGF-beta activated kinase 1/MAP3K7 binding protein 1

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in pH7.3 PBS, 0.05% NaN3, 50% Glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hui Gong et al.
Oncology reports, 39(3), 1090-1098 (2018-01-13)
MicroRNA-873 (miR‑873) has been reported to be dysregulated in a variety of malignancies, however, the biological function and underlying molecular mechanism of miR‑873 in colorectal cancer (CRC) remain unclear. In the present study we found that the expression levels of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service