Skip to Content
Merck
All Photos(1)

Key Documents

SAB4300686

Sigma-Aldrich

Anti-CD3E antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-FLJ18683, Anti-T3E, Anti-TCRE

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$658.00

$658.00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1011


Select a Size

Change View
100 μG
$658.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$658.00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1011

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~21 kDa

species reactivity

mouse, rat, human

concentration

1 mg/mL

technique(s)

western blot: 1:500-1:1000

isotype

IgG

immunogen sequence

(D-L-Y-S-G)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CD3E(916)

General description

The antibody detects endogenous level of total CD3E protein.

Immunogen

Peptide sequence around aa. 197-201 ( D-L-Y-S-G), according to the protein NP_000724.1

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

The CD3 complex mediates signal transduction.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service