Skip to Content
Merck
All Photos(1)

Key Documents

SAB4300657

Sigma-Aldrich

Anti-VEGFR2 (Ab-1059) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-FLK1, Anti-KDR, Anti-VGFR2, Anti-VGR2, Anti-kinase insert domain receptor

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$745.00

$745.00


Check Cart for Availability


Select a Size

Change View
100 μG
$745.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$745.00


Check Cart for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

210-230 kDa

species reactivity

human, mouse

concentration

1 mg/mL

technique(s)

western blot: 1:500-1:1000

isotype

IgG

immunogen sequence

(P-D-Y-V-R)

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... VEGFR2(3791)

General description

The antibody detects endogenous levels of total VEGFR2 protein.

Immunogen

Peptide sequence around aa. 1057-1061 (P-D-Y-V-R), according to the protein NP_002244.1

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

Receptor for VEGF or VEGFC. Has a tyrosine-protein kinase activity. The VEGF-kinase ligand/receptor signaling system plays a key role in vascular development and regulation of vascular permeability. In case of HIV-1 infection, the interaction with extracellular viral Tat protein seems to enhance angiogenesis in Kaposi′s sarcoma lesions

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Praveen Kumar Neeli et al.
The FEBS journal, 290(2), 502-520 (2022-08-27)
DOT1L, a specific H3K79 methyltransferase, has a tumour-promoting role in various cancers, including triple-negative breast cancer (TNBC). However, the molecular mechanism by which the deregulated DOT1L promotes cancer progression is unclear. Herein, we show that a significantly higher basal level

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service