Skip to Content
Merck
All Photos(1)

Key Documents

SAB4300609

Sigma-Aldrich

Anti-FLT4 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-FLT41 antibody produced in rabbit, Anti-LMPH1A antibody produced in rabbit, Anti-PCL antibody produced in rabbit, Anti-VEGFR3 antibody produced in rabbit, Anti-fms-related tyrosine kinase 4 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$512.00

$512.00


Check Cart for Availability


Select a Size

Change View
100 μG
$512.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

$512.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~160 kDa

species reactivity

human

concentration

1 mg/mL

technique(s)

western blot: 1:500-1:1000

isotype

IgG

immunogen sequence

(L-A-S-E-E)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FLT4(2324)

General description

Fms related tyrosine kinase 4 (FLT4) is also known as vascular endothelial growth factor receptor 3 (VEGFR3). It is expressed in the blood and lymphatic endothelial cells during early development. The gene encoding FLT4 is localized on human chromosome 5q35.3.

Immunogen

Peptide sequence around aa. 1279-1283 (L-A-S-E-E), according to the protein FLT4.

Application

Anti-FLT4 antibody produced in rabbit has been used in immunohistochemistry.

Biochem/physiol Actions

Fms related tyrosine kinase 4 (FLT4) has a role in lymphangiogenesis and angiogenesis. It acts as an activator in signaling pathways. Mutations in the gene encoding FLT4 have been linked to Milroy disease.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

Receptor for VEGFC. Has a tyrosine-protein kinase activity.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Two anti-angiogenic TKI-PET tracers, [11C]axitinib and [11C]nintedanib: Radiosynthesis, in vivo metabolism and initial biodistribution studies in rodents
Paul S, et al.
Nuclear Medicine and Biology null
A novel FLT4 Gene mutation and MR lymphangiography in a Chinese family with Milroy disease
Liu, N., et al.
Lymphology, 93-6 (2015)
Molecular controls of lymphatic VEGFR3 signaling
Deng, Yong et al.
Atherosclerosis: Treatment and Prevention (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service