Skip to Content
Merck
All Photos(1)

Key Documents

SAB4300257

Sigma-Aldrich

Anti-phospho-Th (pSer19) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-The antibody produced in rabbit, Anti-tyrosine hydroxylase antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$658.00

$658.00


Estimated to ship on15 May 2025



Select a Size

Change View
100 μG
$658.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$658.00


Estimated to ship on15 May 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

55-60 kDa

species reactivity

human, rat

concentration

1 mg/mL

technique(s)

indirect immunofluorescence: 1:100-1:200

isotype

IgG

immunogen sequence

(A-V-SP-E-Q)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer19)

Gene Information

rat ... Th(25085)

Immunogen

Peptide sequence around phosphorylation site of serine19 (A-V-S(p)-E-Q), according to the protein Th.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

The protein encoded by Tyrosine Hydroxylase is involved in the conversion of tyrosine to dopamine. It is the rate-limiting enzyme in the synthesis of catecholamines, hence plays a key role in the physiology of adrenergic neurons. Mutations in this gene have been associated with autosomal recessive Segawa syndrome. Alternatively spliced transcript variants encoding different isoforms have been noted for this gene.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service