Skip to Content
Merck
All Photos(3)

Key Documents

SAB4300221

Sigma-Aldrich

Anti-phospho-EIF2S1 (pSer51) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-EIF-2 antibody produced in rabbit, Anti-EIF-2A antibody produced in rabbit, Anti-EIF-2alpha antibody produced in rabbit, Anti-EIF2 antibody produced in rabbit, Anti-eukaryotic translation initiation factor 2, subunit 1 alpha, 35kDa antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$745.00

$745.00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1419


Select a Size

Change View
100 μG
$745.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$745.00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1419

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~38 kDa

species reactivity

rat, mouse, human

concentration

1 mg/mL

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50-1:100
indirect immunofluorescence: 1:100-1:200
western blot: 1:500-1:1000

isotype

IgG

immunogen sequence

(E-L-SP-R-R)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer51)

Gene Information

human ... EIF2S1(1965)

Immunogen

Peptide sequence around phosphorylation site of serine 51 (E-L-S(p)-R-R), according to the protein EIF2S1.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

Functions in the early steps of protein synthesis by forming a ternary complex with GTP and initiator tRNA. This complex binds to a 40S ribosomal subunit, followed by mRNA binding to form a 43S preinitiation complex. Junction of the 60S ribosomal subunit to form the 80S initiation complex is preceded by hydrolysis of the GTP bound to eIF-2 and release of an eIF-2-GDP binary complex. In order for eIF-2 to recycle and catalyze another round of initiation, the GDP bound to eIF-2 must exchange with GTP by way of a reaction catalyzed by eIF-2B.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Manuel Bulfoni et al.
Frontiers in endocrinology, 13, 949097-949097 (2022-08-23)
Pancreatic beta cell response to glucose is critical for the maintenance of normoglycemia. A strong transcriptional response was classically described in rodent models but, interestingly, not in human cells. In this study, we exposed human pancreatic beta cells to an
Yilin Yu et al.
Cell metabolism, 29(4), 966-978 (2019-02-19)
Skeletal stem cells (SSCs) are postulated to provide a continuous supply of osteoblasts throughout life. However, under certain conditions, the SSC population can become incorrectly specified or is not maintained, resulting in reduced osteoblast formation, decreased bone mass, and in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service