Skip to Content
Merck
All Photos(1)

Key Documents

SAB4300121

Sigma-Aldrich

Anti-phospho-CHEK2 (pSer516) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-CDS1 antibody produced in rabbit, Anti-CHK2 antibody produced in rabbit, Anti-CHK2 checkpoint homolog (S. pombe) antibody produced in rabbit, Anti-HuCds1 antibody produced in rabbit, Anti-LFS2 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$658.00

$658.00


Check Cart for Availability


Select a Size

Change View
100 μG
$658.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$658.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~62 kDa

species reactivity

human

concentration

1 mg/mL

technique(s)

western blot: 1:500-1:1000

isotype

IgG

immunogen sequence

(Q-P-SP-T-S)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer516)

Gene Information

human ... CHEK2(11200)

Immunogen

Peptide sequence around phosphorylation site of serine 516 (Q-P-S(p)-T-S), according to the protein CHEK2.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

In response to DNA damage and replication blocks, cell cycle progression is halted through the control of critical cell cycle regulators. The protein encoded by Chk2 gene is a cell cycle checkpoint regulator and putative tumor suppressor. It contains a forkhead-associated protein interaction domain essential for activation in response to DNA damage and is rapidly phosphorylated in response to replication blocks and DNA damage. When activated, the encoded protein is known to inhibit CDC25C phosphatase, preventing entry into mitosis, and has been shown to stabilize the tumor suppressor protein p53, leading to cell cycle arrest in G1. In addition, this protein interacts with and phosphorylates BRCA1, allowing BRCA1 to restore survival after DNA damage. Mutations in this gene have been linked with Li-Fraumeni syndrome, a highly penetrant familial cancer phenotype usually associated with inherited mutations in TP53. Also, mutations in this gene are thought to confer a predisposition to sarcomas, breast cancer, and brain tumors. This nuclear protein is a member of the CDS1 subfamily of serine/threonine protein kinases. Three transcript variants encoding different isoforms have been found for this gene.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuanxin Zhang et al.
Oncology letters, 18(2), 1881-1887 (2019-08-20)
Cervical cancer continues to be a threat to female health globally. In the present study, the potential anticancer activity of 2-[2-hydroxyl-1-(4-methoxy phenyl) ethyl]-3-(4-benzyloxy phenyl) isoindolin-1-one (CDS-1548), was evaluated in HeLa cells. CDS-1548 is an organic small-molecule compound characterized by two
Helga B Landsverk et al.
Nucleic acids research, 47(4), 1797-1813 (2018-12-13)
Ataxia telangiectasia mutated and Rad3-related (ATR) kinase is a key factor activated by DNA damage and replication stress. An alternative pathway for ATR activation has been proposed to occur via stalled RNA polymerase II (RNAPII). However, how RNAPII might signal
Shiyuan Hong et al.
Oncogene, 38(17), 3274-3287 (2019-01-12)
High-risk human papillomaviruses (HPVs) constitutively activate ataxia telangiectasia mutated (ATM) and ataxia telangiectasia- and Rad3-related (ATR) DNA damage repair pathways for viral genome amplification. HPVs activate these pathways through the immune regulator STAT-5. For the ATR pathway, STAT-5 increases expression
Bo Liu et al.
The international journal of biochemistry & cell biology, 109, 40-58 (2019-02-03)
The role of protein phosphatase 2ACα (PP2ACα) in brain development is poorly understood. To understand the function of PP2ACα in neurogenesis, we inactivated Pp2acα gene in the central nervous system (CNS) of mice by Cre/LoxP system and generated the PP2ACα
Xingyi Guo et al.
Cancer epidemiology, biomarkers & prevention : a publication of the American Association for Cancer Research, cosponsored by the American Society of Preventive Oncology, 28(8), 1308-1315 (2019-06-05)
Pathogenic variants in susceptibility genes lead to increased breast cancer risk. To identify coding variants associated with breast cancer risk, we conducted whole-exome sequencing in genomic DNA samples from 831 breast cancer cases and 839 controls of Chinese women. We

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service