Skip to Content
Merck
All Photos(3)

Key Documents

SAB4300103

Sigma-Aldrich

Anti-phospho-LIMK1 (pThr508) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-LIM domain kinase 1 antibody produced in rabbit, Anti-LIMK antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$877.00

$877.00


Check Cart for Availability


Select a Size

Change View
100 μG
$877.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$877.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~72 kDa

species reactivity

human, rat, mouse

concentration

1 mg/mL

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50-1:100
indirect immunofluorescence: 1:100-1:200
western blot: 1:500-1:1000

isotype

IgG

immunogen sequence

(R-Y-TP-V-V)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pThr508)

Gene Information

human ... LIMK1(3984)

Immunogen

Peptide sequence around phosphorylation site of threonine 508 (R-Y-T(p)-V-V), according to the protein LIMK1.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

Protein kinase which regulates actin filament dynamics. Phosphorylates and inactivates the actin binding/depolymerizing factor cofilin, thereby stabilizing the actin cytoskeleton. Isoform 3 has a dominant negative effect on actin cytoskeletal changes. May be involved in brain development.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yukie Yamahashi et al.
Molecular psychiatry (2022-06-07)
Acetylcholine is a neuromodulator critical for learning and memory. The cholinesterase inhibitor donepezil increases brain acetylcholine levels and improves Alzheimer's disease (AD)-associated learning disabilities. Acetylcholine activates striatal/nucleus accumbens dopamine receptor D2-expressing medium spiny neurons (D2R-MSNs), which regulate aversive learning through
Dejan Mamula et al.
Journal of leukocyte biology, 110(5), 839-852 (2021-02-03)
Immunity is governed by successful T cell migration, optimized to enable a T cell to fully scan its environment without wasted movement by balancing speed and turning. Here we report that the Arhgef6 RhoGEF (aka alpha-PIX; αPIX; Cool-2), an activator
Lei Zhang et al.
American journal of translational research, 11(7), 4100-4112 (2019-08-10)
Adipose-derived stem cells (ADSCs) are multipotent stromal cells that provide an abundant source of cells for skin tissue engineering and wound healing. Platelet-rich plasma (PRP) is a concentrate of platelet-rich plasma protein, which contains several different growth factors and other
Viviana Guadagni et al.
Scientific reports, 6, 35919-35919 (2016-10-25)
Retinitis pigmentosa (RP) comprises a group of inherited pathologies characterized by progressive photoreceptor degeneration. In rodent models of RP, expression of defective genes and retinal degeneration usually manifest during the first weeks of postnatal life, making it difficult to distinguish

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service