Skip to Content
Merck
All Photos(1)

Key Documents

SAB4300083

Sigma-Aldrich

Anti-phospho-GRIN1 (pSer896) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-NMDA1 antibody produced in rabbit, Anti-NMDAR1 antibody produced in rabbit, Anti-NR1 antibody produced in rabbit, Anti-glutamate receptor, ionotropic, N-methyl D-aspartate 1 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$658.00

$658.00


Check Cart for Availability


Select a Size

Change View
100 μG
$658.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

$658.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~120 kDa

species reactivity

human, mouse, rat

concentration

1 mg/mL

technique(s)

western blot: 1:500-1:1000

isotype

IgG

immunogen sequence

(R-R-SP-S-K)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer896)

Gene Information

human ... GRIN1(2902)

Immunogen

Peptide sequence around phosphorylation site of serine 896 (R-R-S(p)-S-K), according to the protein GRIN1.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

NMDA receptors are members of the ionotropic class of glutamate receptors, which also includes Kainate and AMPA receptors. NMDA receptors consist of NR1 subunits combined with one or more NR2 (A-D) or NR3 (A-B) subunits. The ligand-gated channel is permeable to cations including Ca2+, and at resting membrane potentials NMDA receptors are inactive due to a voltage-dependent blockade of the channel pore by Mg2+. NMDA receptor activation, which requires binding of glutamate and glycine, leads to an influx of Ca2+ into the postsynaptic region where it activates several signaling cascades, including pathways leading to the induction of long-term potentiation (LTP) and depression (LTD). NMDA receptors have a critical role in excitatory synaptic transmission and plasticity in the CNS. They govern a rangeof physiological conditions including neurological disorders caused by excitotoxic neuronal injury, psychiatric disorders and neuropathic pain syndromes.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sho Hasegawa et al.
Neuropharmacology, 148, 107-116 (2018-12-28)
The impairment of social behaviors induced by social defeat stress exposure as juveniles is resistant to some antidepressants and an antipsychotic, although the underlying mechanisms and/or therapeutic target are not yet clear. In this study, we investigated the involvement of
Ming Jiang et al.
Journal of pain research, 12, 2009-2026 (2019-07-17)
Preoperative anxiety is associated with postoperative hyperalgesia; however, few studies have investigated the mechanism underlying this association in female surgical patients. Research has suggested that ON cells in the rostral ventromedial medulla (RVM) receive nerve impulses via cholecystokinin 2 (CCK2)
Bozena E Fyk-Kolodziej et al.
The Journal of comparative neurology, 529(9), 2311-2331 (2020-12-22)
The rostral ventrolateral medulla (RVLM) is a brain region involved in normal regulation of the cardiovascular system and heightened sympathoexcitatory states of cardiovascular disease (CVD). Among major risk factors for CVD, sedentary lifestyles contribute to higher mortality than other modifiable
Huijie Ma et al.
The Journal of physiology, 597(16), 4325-4340 (2019-06-27)
The angiotensin AT1 receptor expression and protein kinase C (PKC)-mediated NMDA receptor phosphorylation levels in the hypothalamus are increased in a rat genetic model of hypertension. Blocking AT1 receptors or PKC activity normalizes the increased pre- and postsynaptic NMDA receptor

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service