Skip to Content
Merck
All Photos(2)

Key Documents

SAB4300038

Sigma-Aldrich

Anti-phospho-STAT6 (pTyr641) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-D12S1644 antibody produced in rabbit, Anti-IL-4-STAT antibody produced in rabbit, Anti-STAT6B antibody produced in rabbit, Anti-STAT6C antibody produced in rabbit, Anti-signal transducer and activator of transcription 6, interleukin-4 induced antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$877.00

$877.00


Estimated to ship on01 May 2025



Select a Size

Change View
100 μG
$877.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

$877.00


Estimated to ship on01 May 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~110 kDa

species reactivity

human

concentration

1 mg/mL

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50-1:100
western blot: 1:500-1:1000

isotype

IgG

immunogen sequence

(R-G-YP-V-P)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pTyr641)

Gene Information

human ... STAT6(6778)

Immunogen

Peptide sequence around phosphorylation site of tyrosine 641 (R-G-Y(p)-V-P), according to the protein STAT6.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

Carries out a dual function: signal transduction and activation of transcription. Involved in interleukin-4 signalling.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marshall E Kadin et al.
Aesthetic surgery journal, 43(2), 137-146 (2022-08-25)
Breast implant-associated anaplastic large cell lymphoma (BIA-ALCL) is a rare, usually indolent CD30+ T-cell lymphoma with tumor cells, often surrounded by eosinophils, expressing IL-13 and pSTAT6. The aim of this study was to understand the unique tumor pathology and growth
Alfred Hj Kim et al.
JCI insight, 2(21) (2017-11-03)
The efficacy of B cell depletion therapies in diseases such as nephrotic syndrome and rheumatoid arthritis suggests a broader role in B cells in human disease than previously recognized. In some of these diseases, such as the minimal change disease

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service