Skip to Content
Merck
All Photos(2)

Key Documents

SAB4300031

Sigma-Aldrich

Anti-phospho-GATA1 (pSer310) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-ERYF1 antibody produced in rabbit, Anti-GATA binding protein 1 (globin transcription factor 1) antibody produced in rabbit, Anti-GF-1 antibody produced in rabbit, Anti-GF1 antibody produced in rabbit, Anti-NFE1 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~43 kDa

species reactivity

rat, mouse, human

concentration

1 mg/mL

technique(s)

indirect immunofluorescence: 1:100-1:200
western blot: 1:500-1:1000

isotype

IgG

immunogen sequence

(K-A-SP-G-K)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer310)

Gene Information

human ... GATA1(2623)

General description

GATA binding protein 1 (GATA1) is a key hematopoietic transcription factor. It is encoded by the gene mapped to human chromosome Xp11.23 and it belongs to the GATA transcription factor family.

Immunogen

Peptide sequence around phosphorylation site of serine 310 (K-A-S(p)-G-K), according to the protein GATA1.

Biochem/physiol Actions

GATA binding protein 1 (GATA1) plays a crucial role in erythroid and megakaryocyte differentiation and proliferation. It modulates the expression of genes and acts as an E-cadherin repressor. Mutations in the GATA1 gene have been linked to transient abnormal myelopoiesis (TAM) and myeloid leukemia of DS (ML-DS). The protein has anti-apoptotic functions in breast cancer.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

GATA1 encodes a protein which belongs to the GATA family of transcription factors. The protein plays an important role in erythroid development by regulating the switch of fetal hemoglobin to adult hemoglobin. Mutations in this gene have been associated with X-linked dyserythropoietic anemia and thrombocytopenia.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

GATA1 mutations in a cohort of Malaysian children with Down syndrome-associated myeloid disorder.
Lum SH
Singapore Medical Journal, 57, 320-324 (2016)
GATA1 induces epithelial-mesenchymal transition in breast cancer cells through PAK5 oncogenic signaling.
Li Y
Oncotarget, 6, 4345-4356 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service