Skip to Content
Merck
All Photos(2)

Key Documents

SAB4200859

Sigma-Aldrich

Anti-Staphylococcus Enterotoxin B antibody, Mouse monoclonal

clone SEB4, purified from hybridoma cell culture

Synonym(s):

Enterotoxin type B, SEB

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$302.00
100 μL
$765.00

$302.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μL
$302.00
100 μL
$765.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$302.00


Check Cart for Availability

Request a Bulk Order

antibody form

purified from hybridoma cell culture

Quality Level

clone

SEB4

form

liquid

species reactivity

Staphylococcus aureus ((Staphylococcal enterotoxin B))

concentration

~1 mg/mL

technique(s)

ELISA: 0.12-0.25 μg/mL using 1 μg/ml staphylococcal enterotoxin B for coating.
immunoblotting: 0.5-1 μg/mL using staphylococcal enterotoxin B.

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Staphylococcus aureus is a Gram-positive bacterium, that causes disease in the human population including food poisoning and toxic shock syndrome.1 S. aureus expresses several secreted virulence factors, such as various enzymes, cytotoxins, exotoxins, and exfoliative toxins.1 Exotoxins include more than 20 serologically classified staphylococcal enterotoxins (SEs)

Specificity

Monoclonal Anti-Staphylococcal Enterotoxin B antibody specifically recognizes staphylococcal enterotoxin B (SEB) and has no cross reactivity with staphylococcal enterotoxin A (SEA), cholera toxin, or pseudomonas exotoxin A.

Application

The antibody may be used in various immunochemical techniques including Immunoblotting and ELISA. Detection of the staphylococcal enterotoxin B bands by Immunoblotting is specifically inhibited by the immunogen.

Biochem/physiol Actions

Exotoxins include more than 20 serologically classified staphylococcal enterotoxins (SEs). The best characterized are the SEs A, B through V, and toxic shock syndrome toxin-1 (TSST-1). These enterotoxins are similar in activity, sequence, structure, and molecular mass (25-30 kDa).1-3 SEA and SEB are known as superantigens due to their ability to bind class II MHC molecules on antigen presenting cells. This binding stimulates extensive T-cell activation followed by massive cytokine release leading to an acute toxic shock.1-4 SEB is also classified as a category B select agent and is important toxicant in biodefense research.5-9SE proteins have a significant resistance to heat and acid, and are also resistant to gastrointestinal proteases including pepsin, trypsin, rennin, and papain. Thus, inactivating the S. aureus bacteria may not be sufficient to eliminate the risk of its superantigens from causing food poisoning.1SEB contamination can be caused by both its ingestion (resulting with food poisoning) or by inhalation (resulting with respiratory symptoms of cough and dyspnea), therefore SEB detection is a major challenge in the food industry.1,3,5-10

Physical form

Supplied as a solution in 0.01 M phosphate buffered saline pH 7.4, containing 15 mM sodium azide as a preservative.

Storage and Stability

For continuous use, store at 2-8°C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog  our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service