Skip to Content
Merck
All Photos(2)

Key Documents

SAB4200844

Sigma-Aldrich

Anti-Cholera toxin, B Subunit (CTxB) antibody, Mouse monoclonal

clone CTxB-24, purified from hybridoma cell culture

Synonym(s):

Anti-toxB

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$743.00

$743.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μL
$743.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$743.00


Check Cart for Availability

Request a Bulk Order

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

CTxB-24, monoclonal

description

Research area: Microbiome

species reactivity

Vibrio cholerae

packaging

antibody small pack of 25 μL

concentration

~1 mg/mL

technique(s)

immunoblotting: 1-2 μg/mL using His-tagged recombinant Cholera Toxin B subunit

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Related Categories

General description

Cholera toxin (CTx) also known as choleragen, is an enterotoxin produced by the Gram-negative bacterium Vibrio cholerae that naturally habitat in fresh or saltwater environments. Most of the V. cholerae species do not cause any disease in human, but few including serotypes O1 and O139 can cause cholera pandemic. These cases were described already in the 19th century.1 The V. cholerae virulence factors CtxA and CtxB are located at the CTX phage genome integrated within the bacterial chromosome. Since species virulence may change due to mutations and acquisition of virulence genes, the cholera pandemic has a major public health risk with potential for large numbers of cases and even deaths.1-3

Specificity

Monoclonal Anti-Cholera Toxin-peroxidase antibody specifically recognizes Cholera Toxin and has no cross reactivity with Staphylococcal Enterotoxin A (SEA), Staphylococcal Enterotoxin B (SEB), Pseudomonas Exotoxin A or Staphylococcal Alpha-Toxin (α-Hemolysin).

Immunogen

recombinant Cholera toxin beta (C-terminal His-tag) protein.

Application

The antibody may be used in various immunochemical techniques including Immunoblotting, Immunofluorescence and ELISA. Detection of the CTxB band by Immunoblotting is specifically inhibited by the immunogen.

Biochem/physiol Actions

CTx is composed of two subunits, the toxic CTxA (~27 kDa) and non-toxic CTxB (~12 kDa) assembled with the stoichiometry AB5.4 The B-subunit specifically binds to monosialogangliosides GM1 receptors, located in the membrane of intestinal epithelial cells.5 The A1 fragment of the A-subunit is translocated through the membrane of the host cell, where it catalyses the ADP-ribosylation of the Gsa regulatory component of the adenylate cyclase complex. The resulting increased level of cyclic AMP promotes a wide variety of actions, including the secretion of chloride ions in the case of intestinal epithelial cells.6-7 Antibodies specific for cholera toxin may be used in studies of structural and functional aspects of toxin-membrane interactions and for the detection of CTxB when used for example as an adjuvant when injected mucosally together with the desired antigen.8-10

Physical form

Supplied as a solution in 0.01 M phosphate buffered saline pH 7.4, containing 15 mM sodium azide as a preservative.

Storage and Stability

For continuous use, store at 2-8°C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sumio Hayakawa et al.
JCI insight, 7(22) (2022-12-13)
Recent studies have shown that cellular metabolism is tightly linked to the regulation of immune cells. Here, we show that activation of cholesterol metabolism, involving cholesterol uptake, synthesis, and autophagy/lipophagy, is integral to innate immune responses in macrophages. In particular
Hui Gong et al.
Cellular and molecular life sciences : CMLS, 81(1), 39-39 (2024-01-12)
Colorectal cancer (CRC) is characterized by a complex tumor inflammatory microenvironment, while angiogenesis and immunosuppression frequently occur concomitantly. However, the exact mechanism that controls angiogenesis and immunosuppression in CRC microenvironment remains unclear. Herein, we found that expression levels of lipid

Questions

  1. May I know the fairly secondary antibody for the CtxB primary antibody?

    1 answer
    1. When selecting a secondary antibody to react with the primary mouse antibody, one may want to choose an anti-mouse IgG antibody compatible with the desired experimental applications, e.g., Western blot (WB), immunohistochemistry (IHC), enzyme-linked immunosorbent assay (ELISA), or immunofluorescence (IF). The following representative products may be of consideration.
      SAB3701171 Anti-Mouse IgG1 (γ-chain specific)-Peroxidase antibody produced in rabbit
      F0257 Anti-Mouse IgG (whole molecule)–FITC antibody produced in goat affinity isolated antibody

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service