Skip to Content
Merck
All Photos(1)

Key Documents

SAB4200787

Sigma-Aldrich

Anti-Secretory Component (IgA) antibody, Mouse monoclonal

clone GA-1, purified from hybridoma cell culture

Synonym(s):

Anti-Secretory IgA

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$759.00

$759.00


Available to ship on28 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μL
$759.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

$759.00


Available to ship on28 April 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

GA-1, monoclonal

form

buffered aqueous solution

species reactivity

human

concentration

~1 mg/mL

technique(s)

indirect ELISA: 0.06-0.12 μg/mL using 5 μg/ml secretory human IgA for coating.

isotype

IgG1

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

The secretory component is a single chain glycoprotein which is synthesized principally by epithelial cells in mucous membranes and exocrine glands. It occurs both in a free form and as a subunit of the secretory immunoglobulin A (SIgA) molecule. It has a molecular weight of approximately 75,000 Daltons. The secretory component is attached to the immunoglobulin molecule during the secretion process. Its biological function has not been established although several possibilities for which there are varying degrees of support have been suggested. These possibilities include: protection of IgA against destruction by proteolysis, transport of IgA across the epithelial surface and attraction of circulating lymphocytes with surface IgA to mucous membranes. Secretory IgA is also present in circulating blood and high concentrations of SIgA or free secretory component are reported in patients with carcinomas and chronic infectious diseases. Human IgA accounts for approximately 20% of all immunoglobulins found in adult human serum. It consists of two heavy chains and two light chains. In serum it is usually found as monomeric but in secretions it exists as a dimer linked by a J-chain and associated by a peptide secretory component. Although IgA has been shown to have the usual antibody properties it is probably more important in secretions (saliva, colostrum, tears, nasal, bronchial and intestinal) where it has the role of creating an immune barrier against various microorganisms at exposedmucous surfaces. Monoclonal Anti-Secretory Component (IgA) may be used for quantitative determination of human secretory component or secretory IgA in various body fluids and immunohistochemical localization of secretory component in mucous membrane tissue.

Immunogen

Secretory component purified from human colostrum.

Application

Indirect ELISA: a working concentration of 0.06-0.12 μg/mL is recommended using 5 μg/mL secretory human IgA for coating.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15mM sodium azide.

Other Notes

In order to obtain best results in different techniques and preparations we recommend determining optimal working concentration by titration test.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kathrin Göritzer et al.
International journal of molecular sciences, 25(13) (2024-07-13)
Secretory IgA (SIgA) presents a promising avenue for mucosal immunotherapy yet faces challenges in expression, purification, and stability. IgA exists in two primary isotypes, IgA1 and IgA2, with IgA2 further subdivided into two common allotypes: IgA2m(1) and IgA2m(2). The major

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service