Skip to Content
Merck
All Photos(3)

Key Documents

SAB4200710

Sigma-Aldrich

Anti-Calponin antibody, Mouse monoclonal

clone hCP, purified from hybridoma cell culture

Synonym(s):

Anti-CNN-1, Anti-CNN-2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$830.00

$830.00


Check Cart for Availability


Select a Size

Change View
100 μL
$830.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$830.00


Check Cart for Availability

antibody form

purified from hybridoma cell culture

Quality Level

antibody product type

primary antibodies

clone

hCP, monoclonal

form

buffered aqueous solution

mol wt

antigen ~34 kDa

species reactivity

bovine, monkey, human, rabbit, ferret, mouse, hamster, rat, canine, pig

concentration

~1 mg/mL

technique(s)

immunoblotting: suitable
immunohistochemistry: 10-20 μg/mL using heat-retrieved formalin-fixed, paraffin-embedded human tonsil sections
immunoprecipitation (IP): suitable

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CNN1(1264)

General description

Anti-Calponin antibody, Mouse monoclonal (mouse IgG1 isotype) is derived from the hCP hybridoma (also cites as CALP), produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mouse. Calponin (CNN) exists in three isoforms CNN1, CNN2 and CNN3. CNN2 gene is mapped to human chromosome 19p13.3. All three isoforms are expressed in smooth muscle. While CNN1 expression is confined only to smooth muscle, CNN2 and CNN3 are expressed in other cells including the neuronal tissue, fibroblast, myoblast and B lymphocyte. The three isoforms differ in the C-terminal region and comprise conserved actin-binding middle domain and a N-terminal calponin homology (CH) domain.

Specificity

Monoclonal Anti-Calponin specifically recognizes smooth muscle Calponin.

Immunogen

human uterus smooth muscle extract

Application

Anti-Calponin antibody, Mouse monoclonal may be used in:
  • immunoblotting
  • immunoprecipitation
  • immunohistochemistry

Biochem/physiol Actions

Calponin is an F-actin, calmodulin, and tropomyosin binding protein that presents in both the cytoskeleton and contractile machinery of smooth muscle (SM) cells. The interaction of Calponin with actin is shown to inhibit myosin Mg-ATPase activity. The binding strength of calponin to tropomyosin is weakened in the presence of Ca2+. Calponin is involved in a variety of cellular processes including smooth muscle contraction, cell movement, shape change, angiogenesis, exocytosis and endocytosis. Monoclonal anti-Calponin can be used for the identification and localization of calponin in SM elements. Calponin levels may serve as a prognostic factor in hepatocellular carcinoma, renal cell carcinoma, osteosarcoma and melanoma.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8°C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Calcium-dependent regulation of smooth muscle calponin by S100
Fujii T, et al.
Journal of Biochemistry, 116(1), 121-127 (1994)
Calponin isoforms CNN1, CNN2 and CNN3: regulators for actin cytoskeleton functions in smooth muscle and non-muscle cells
Liu R and Jin JP
Gene, 585(1), 143-153 (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service