Skip to Content
Merck
All Photos(3)

Key Documents

SAB4200617

Sigma-Aldrich

Monoclonal Anti-Myb antibody from mouse

clone MB-8, purified immunoglobulin

Synonym(s):

Proto-oncogene c-Myb, Transcriptional activator Myb, Transcriptional activator Myb; Proto-oncogene c-Myb

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
$621.00

$621.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
200 μL
$621.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$621.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

MB-8, monoclonal

species reactivity

human, mouse

concentration

~1 mg/mL

technique(s)

flow cytometry: 2.5-5 μg/test using using HL-60 cells.
immunoblotting: 0.5-1 μg/mL using using Jurkat total cell extract.
immunofluorescence: 2-4 μg/mL using using HeLa cells.

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MYB(4602)

General description

Monoclonal Anti-Myb (mouse IgG1 isotype) is derived from the hybridoma MB-8 produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice. MYB proto-oncogene, transcription factor is encoded by the gene mapped to human chromosome 6q23.3.

Specificity

Monoclonal Anti-Myb recognizes human and mouse Myb.

Immunogen

synthetic peptide corresponding to a sequence at the N-terminal region of human Myb.

Application

Monoclonal Anti-Myb antibody from mouse may be used in:
  • immunoblotting
  • flow cytometry
  • immunocytochemistry

Biochem/physiol Actions

MYB is a transcription factor that regulates stem and progenitor cells in the bone marrow, colonic crypts and a neurogenic region of the adult brain. Loss of MYB activity in these cells leads to severe or lethal phenotypes. MYB is an oncogene and is observed in some human leukemias. In addition, It is also observed that MYB is activated in colon and breast cancer.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For extended storage, freeze at -20 ˚C in working aliquots. Repeated freezing and thawing, or storage in “frost-free” freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The HBS1L-MYB intergenic region on chromosome 6q23. 3 influences erythrocyte, platelet, and monocyte counts in humans
Menzel S, et al.
Blood, 110(10), 3624-3626 (2007)
MYB function in normal and cancer cells
Ramsay RG, et al.
Nature Reviews. Cancer, 8, 523-534 (2008)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service