Skip to Content
Merck
All Photos(3)

Key Documents

SAB4200615

Sigma-Aldrich

Monoclonal Anti-WDR49 antibody produced in mouse

~1.0 mg/mL, clone W49.1, purified immunoglobulin

Synonym(s):

Anti-WD repeat-containing protein 49, Anti-WDR49

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

W49.1, monoclonal

form

buffered aqueous solution

mol wt

antigen ~85 kDa

species reactivity

human

concentration

~1.0 mg/mL

technique(s)

flow cytometry: 5-10 μg/test using Jurkat cells
immunoblotting: 20 μg/mL using total cell extracts of HEK-293T cells overexpressing WDR49
immunocytochemistry: suitable
immunofluorescence: 10-20 μg/mL using HeLa cells

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... WDR49(151790)

General description

Monoclonal Anti-WDR49 (mouse IgG1 isotype) is derived from the hybridoma W49.1 produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with a synthetic peptide. WD repeat-containing protein 49 (WDR49) is expressed as two isoforms. It is located on the human chromosome at 3q26.1.

Specificity

Monoclonal Anti-WDR49 recognizes human WDR49.

Immunogen

synthetic peptide corresponding to an internal sequence of human WDR49

Application

Monoclonal Anti-WDR49 antibody produced in mouse may be used in:
  • immunoblotting
  • flow cytometry
  • immunocytochemistry
  • immunofluorescence

Biochem/physiol Actions

WD repeat-containing protein 49 (WDR49) is deleted in liver cancer as well as fatal, disomy 3 uveal melanoma. It may play a role as a tumor suppressor gene.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For extended storage, freeze at -20 °C in working aliquots. Repeated freezing and thawing,orstorage in “frost-free” freezers,is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sex-Specific Transcriptome Differences in Substantia Nigra Tissue: A Meta-Analysis of Parkinson?s Disease Data
Mariani E, et al.
Genes, 9(6), 275-275 (2018)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service