Skip to Content
Merck
All Photos(2)

Key Documents

SAB4200566

Sigma-Aldrich

Anti-KHSRP antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody

Synonym(s):

Anti-FBP2, Anti-FUBP2, Anti-KH-type splicing regulatory protein, Anti-KSRP, Anti-MGC99676

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
$528.00

$528.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
200 μL
$528.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$528.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~82 kDa

species reactivity

rat, mouse, human

concentration

~1.0 mg/mL

technique(s)

immunoprecipitation (IP): 1-2 μg using lysates of human HEK-293T cells
western blot: 0.25-0.50 μg/mL using whole extracts of mouse NIH-3T3 cells

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KHSRP(8570)
mouse ... Khsrp(16549)
rat ... Khsrp(171137)

Looking for similar products? Visit Product Comparison Guide

General description

K-homology type-splicing regulatory protein (KHSRP) is a multifunctional RNA-binding protein. It contains four K homology RNA-binding domains and an unusual C-terminal domain. KHSRP is localized in neural and non-neural cell lines. It is located on the human chromosome at 19p13.3.

Specificity

Anti-KHSRP recognizes human, mouse and rat KHSRP.

Immunogen

synthetic peptide corresponding to an internal region of human KHSRP (GeneID: 8570), conjugated to KLH. The corresponding sequence is identical in rat, mouse, monkey, pig and dog KHSPR.

Application

Anti-KHSRP antibody produced in rabbit may be used in immunoblotting and immunoprecipitation.

Biochem/physiol Actions

K-homology type-splicing regulatory protein (KHSRP) is involved in several cellular processes such as transcription, mRNA localization and alternative pre-mRNA splicing. It is also involved in mRNA decay and microRNA precursor maturation.

Physical form

Solution in 0.01 M phosphate buffered saline pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage freeze in working aliquots. Repeated freezing and thawing is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A new regulatory protein, KSRP, mediates exon inclusion through an intronic splicing enhancer.
Min H, et al.
Genes & Development, 11(8), 1023-1036 (1997)
Diverse roles of the nucleic acid-binding protein KHSRP in cell differentiation and disease
Briata P, et al.
Wiley Interdisciplinary Reviews. RNA, 7(2), 227-240 (2016)
Mapping of theKHSRPGene to a Region of Conserved Synteny on Human Chromosome 19p13. 3 and Mouse Chromosome 17
Ring H Z, et al.
Genomics, 56(3), 350-352 (1999)
A Membraneless Organelle Associated with the Endoplasmic Reticulum Enables 3'UTR-Mediated Protein-Protein Interactions
Ma W and Mayr C
Cell, 175(6), 1492-1506 (2018)
Gain of Additional BIRC3 Protein Functions through xn-3-t6a-UTR-Mediated Protein Complex Formation
Lee SH and Mayr C
Molecular Cell, 74(4), 701-712 (2019)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service