Skip to Content
Merck
All Photos(2)

Key Documents

SAB4200358

Sigma-Aldrich

Anti-PLAUR antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody

Synonym(s):

Anti-CD87, Anti-U-PAR, Anti-UPAR, Anti-URKR, Anti-Urokinase receptor, Anti-plasminogen activator

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 45/50 kDa

species reactivity

human

concentration

~1.0 mg/mL

technique(s)

indirect immunofluorescence: 1-2 μg/mL using human HeLa cells.
western blot: 0.5-1.0 μg/mL using whole extracts of HEK-293T cells over-expressing human PLAUR.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PLAUR(5329)

General description

Plasminogen activator urokinase receptor (PLAUR)/ urokinase plasminogen activator receptor (uPAR), a member of Ly6/ neurotoxin receptor family, is a heavily glycosylated glycosyl-phosphatidylinositol (GPI)-anchored cell-surface receptor. It is comprised of serine protease uPA, specific inhibitors (plasminogen activator inhibitors 1 and 2) and three internally disulfide-bonded domains (D1, D2 and D3).

Immunogen

peptide corresponding to a sequence at the C-terminus of human PLAUR, conjugated to KLH. The corresponding sequence is identical in monkey PLAUR.

Application

Anti-PLAUR antibody produced in rabbit has been used in immunoblotting and immunofluorescence.

Biochem/physiol Actions

Plasminogen activator urokinase receptor (PLAUR)/ urokinase plasminogen activator receptor (uPAR) acts as a receptor for urokinase plasminogen activator (uPA). It localizes and promotes plasmin formation. uPAR influences many normal and pathological processes related to cell surface plasminogen activation, degradation of the extracellular matrix, cell migration, leukocyte adhesion, chemotaxis and signal transduction during leukocyte recruitment from the circulation to extravascular sites of inflammation. It is implicated as an anti-cancer target. uPAR is involved in epilepsy, language, cognition, communication and central nervous system disorders.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

uPAR as anti-cancer target: evaluation of biomarker potential, histological localization, and antibody-based therapy
K Lund I, et al.
Current Drug Targets, 12(12), 1744-1760 (2011)
The role of the urokinase receptor in epilepsy, in disorders of language, cognition, communication and behavior, and in the central nervous system
Bruneau N and Szepetowski P
Current Pharmaceutical Design, 17(19), 1914-1923 (2011)
CD87 (urokinase-type plasminogen activator receptor), function and pathology in hematological disorders: a review
Bene MC, et al.
Leukemia, 18(3), 394-394 (2004)
Urokinase plasminogen activator receptor choreographs multiple ligand interactions: implications for tumor progression and therapy
Mazar AP
Clinical Cancer Research, 14(18), 5649-5655 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service